Skip to Content
Merck
All Photos(1)

Key Documents

73837

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl α-D-glucopyranoside

≥98.0% (HPLC), powder

Synonym(s):

X-α-Glc, X-α-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

5-Bromo-4-chloro-3-indolyl α-D-glucopyranoside, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D 105.0±5.0°, c = 1 in DMF/H2O 1:1

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11-,12+,13-,14+/m1/s1

InChI key

OPIFSICVWOWJMJ-HRNXZZBMSA-N

General description

5-Bromo-4-chloro-3-indolyl α-D-glucopyranoside, X-Gluc, is a indigogenic substrate for β- glucuronidase enzyme (GUS). It aids screening of transgenic plants based on GUS activity. GUS deesterifies X-Gluc into indoxyl derivative which upon oxidative polymerization results in the generation of blue indigotin dye.

Application

suitable as substrate for α-glucosidase; substrate used in the Druggan-Forsythe-Iversen medium (DFI agar) for the selective detection of Enterobacter sakazakii in infant milk

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GUS fusions: beta-glucuronidase as a sensitive and versatile gene fusion marker in higher plants.
Jefferson RA, et al.
The Embo Journal, 6(13), 3901-3907 (1987)
Carol Iversen et al.
International journal of food microbiology, 96(2), 133-139 (2004-09-15)
Enterobacter sakazakii can cause fatal invasive infection of neonates associated with the presence of this organism in powdered infant milk formula. A new chromogenic medium (Druggan-Forsythe-Iversen agar, DFI) is described for the selective detection of this emergent pathogen. The medium
B T Kinsella et al.
Current genetics, 20(1-2), 45-52 (1991-07-01)
The molecular cloning of an alpha-glucosidase gene isolated from a Candida tsukubaensis (CBS 6389) genomic library in Saccharomyces cervisiae is reported. The cloned gene is contained within a 6.2 kb Sau3A DNA fragment and directs the synthesis and secretion of
A Histochemical Method Using a Substrate of. BETA.-Glucuronidase for Detection of Genetically Modified Papaya
Wakui C, et al.
Shokuhin Eiseigaku Zasshi, 45(1), 19-24 (2004)
Functional insight for beta-glucuronidase in Escherichia coli and Staphylococcus sp. RLH1
Arul L, et al.
Bioinformation, 2(8), 339-339 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service