Skip to Content
Merck
All Photos(3)

Documents

B9146

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside

≥98%, powder, suitable for identification of lac+ bacterial colonies

Synonym(s):

5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein:
1402009
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, ≥98%

Assay

≥98%

form

powder

solubility

DMF: methanol (1:1): soluble 50 mg/mL

suitability

suitable for identification of lac+ bacterial colonies

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11+,12+,13-,14-/m1/s1

InChI key

OPIFSICVWOWJMJ-AEOCFKNESA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside has been used to confirm transduction of Caki-2/LacZ clones. It has also been used in blue-white selection of transformants.
X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

Reconstitution

A stock solution (100 mg/ml) is prepared by dissolving X-Gal in dimethylformamide and should be stored at −20 °C and protected from light. Sterilization is not required.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HSV-tk gene therapy for human renal cell carcinoma in nude mice
Pulkkanen K, et al.
Cancer Gene Therapy, 8(7), 529-529 (2001)
Rolling circle amplification to screen yam germplasm for badnavirus infections and to amplify and characterise novel badnavirus genomes
Bomer M, et al.
Bio-protocol, 8(1), 529-529 (2018)
Yibing Wang et al.
PloS one, 8(3), e59195-e59195 (2013-03-26)
Our study had three objectives: to extend the plasmid-based transformation protocol to a clinical isolate of C. trachomatis belonging to the trachoma biovar, to provide "proof of principle" that it is possible to "knock out" selected plasmid genes (retaining a
Amy McCurley et al.
Nature medicine, 18(9), 1429-1433 (2012-08-28)
Hypertension is a cardiovascular risk factor present in over two-thirds of people over age 60 in North America; elevated blood pressure correlates with increased risk of heart attack, stroke and progression to heart and kidney failure. Current therapies are insufficient
Florian Weinberger et al.
Circulation research, 110(10), 1303-1310 (2012-03-20)
The transcription factor Islet-1 is a marker of cardiovascular progenitors during embryogenesis. The isolation of Islet-1-positive (Islet-1(+)) cells from early postnatal hearts suggested that Islet-1 also marks cardiac progenitors in adult life. We investigated the distribution and identity of Islet-1(+)

Articles

General protocols for growth of competent cells and their transformation (uptake of DNA).

General protocols for growth of competent cells and their transformation (uptake of DNA).

General protocols for growth of competent cells and their transformation (uptake of DNA).

General protocols for growth of competent cells and their transformation (uptake of DNA).

See All

Protocols

Technical Article on competent cells. Transformation is a process by which some bacteria take up foreign genetic material (naked DNA) from the environment.

Technical Article on competent cells. Transformation is a process by which some bacteria take up foreign genetic material (naked DNA) from the environment.

Technical Article on competent cells. Transformation is a process by which some bacteria take up foreign genetic material (naked DNA) from the environment.

Technical Article on competent cells. Transformation is a process by which some bacteria take up foreign genetic material (naked DNA) from the environment.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service