Skip to Content
Merck
All Photos(1)

Documents

68903

Sigma-Aldrich

4-Methylumbelliferyl β-D-galactopyranoside-6-sulfate sodium salt

galactose-6-sulfate sulphatase substrate, ≥90% (HPLC), powder

Synonym(s):

4-MU-Gal-6S sodium salt, 4-Methylumbelliferyl Beta-D-galactopyranoside-6-sulfate sodium salt, 4-Methylumbelliferyl Gal-6S sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NaO11S
CAS Number:
Molecular Weight:
440.35
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

4-Methylumbelliferyl β-D-galactopyranoside-6-sulfate sodium salt, ≥90% (HPLC)

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Na+].CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H](COS([O-])(=O)=O)[C@H](O)[C@H](O)[C@H]3O)ccc12

InChI

1S/C16H18O11S.Na/c1-7-4-12(17)26-10-5-8(2-3-9(7)10)25-16-15(20)14(19)13(18)11(27-16)6-24-28(21,22)23;/h2-5,11,13-16,18-20H,6H2,1H3,(H,21,22,23);/q;+1/p-1/t11-,13+,14+,15-,16-;/m1./s1

InChI key

KHTOBGQRIASILC-XNSOHGFJSA-M

Application

suitable as a fluorogenic substrate for galactose-6-sulfate sulphatase; fluorogenic substrate for the diagnosis of Morquio disease type A

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Zhao et al.
Prenatal diagnosis, 10(2), 85-91 (1990-02-01)
A new fluorogenic substrate, 4 methylumbelliferyl beta-D-6-sulphogalactoside, was used for the assay of galactose-6-sulphate sulphatase activity in chorionic villi, cultured villus cells, and amniocytes. The fluorometric assay is much more convenient than the conventional assay using radiolabelled, sulphated oligosaccharides. Both
O P van Diggelen et al.
Clinica chimica acta; international journal of clinical chemistry, 187(2), 131-139 (1990-02-28)
4-Methylumbelliferyl-beta-D-galactopyranoside-6-sulphate was synthesized and used for the determination of galactose-6-sulphate sulphatase activity. Fibroblasts and leucocytes from 12 different Morquio A patients, showed 0.0-2.7% of mean normal galactose-6-sulphate sulphatase activity. Heterozygotes showed intermediate activities. The enzymatic liberation of the fluorochrome from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service