Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
egg yolk
Quality Segment
form
solid
concentration
~60% (TLC)
functional group
phospholipid
lipid type
phosphoglycerides
shipped in
ambient
storage temp.
−20°C
SMILES string
[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C
InChI
1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1
InChI key
JLPULHDHAOZNQI-ZTIMHPMXSA-N
Application
- Epithelial transport and bioavailability of intranasally administered human growth hormone formulated with the absorption enhancers didecanoyl-L-alpha-phosphatidylcholine and alpha-cyclodextrin in rabbits.: This research explores the bioavailability and epithelial transport of intranasally administered human growth hormone, highlighting the role of phosphatidylcholine as an absorption enhancer. (Agerholm et al., 1994).
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| description egg yolk, ~60% (TLC) | description egg yolk, Type XIII-E, 100 mg/mL in ethanol, solution | description egg yolk, Type XI-E, 100 mg/mL in chloroform, ≥99%, solution | description from soybean, Type II-S, 14-29% choline basis |
| biological source egg yolk | biological source egg yolk | biological source egg yolk | biological source soybean |
| functional group phospholipid | functional group phospholipid | functional group phospholipid | functional group phospholipid |
| form solid | form solution | form solution | form powder or solid |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| shipped in ambient | shipped in - | shipped in - | shipped in ambient |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Still not finding the right product?
Explore all of our products under L-α-Phosphatidylcholine
Storage Class
11 - Combustible Solids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
