Skip to Content
Merck
All Photos(1)

Key Documents

60238

Sigma-Aldrich

Potassium fluoride

BioUltra, ≥99.5% (F)

Synonym(s):

KF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
Beilstein:
3902818
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent grade
form:
solid
solubility:
H2O: 1 M at 20 °C, clear, colorless

grade

reagent grade

Quality Level

product line

BioUltra

Assay

≥99.5% (F)

form

solid

impurities

insoluble matter, passes filter test
≤0.01% free acid (as HF)
≤0.01% free alkali (as KOH)
≤0.1% potassium fluoro silicate (K2SiF6)

pH

7.0-8.5 (25 °C, 1 M in H2O)

mp

858 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤3 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤2000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.
Potassium fluoride may be used in thermodynamic analysis of DNA-ligand binding and studies on the structural stability of proteins and peptides.

Other Notes

Inhibitor of pyrophosphatase, hence prevents loss of ATP by hydrolytic cleavage of the pyrophosphate bond in the assay of amidophosphoribosyltransferase; Strongly inhibits polypeptide chain initiation in the reticulocyte lysate cell-free system

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J C O'Rourke et al.
The Journal of biological chemistry, 250(9), 3443-3450 (1975-05-20)
KF (30 mM) strongly inhibits polypeptide chain initiation in the reticulocyte lysate cell-free system.. Chain elongation is partially inhibited but proceeds to a significant extent with little initiation of new chains. Polysome breakdown is incomplete after incubations as long as
Amidophosphoribosyltransferase (chicken liver).
J M Lewis et al.
Methods in enzymology, 51, 171-178 (1978-01-01)
Mingming Fan et al.
Bioresource technology, 104, 447-450 (2011-12-16)
Transesterification of soybean oil with methanol was carried out in the presence of CaO-MgO and KF-modified CaO-MgO catalysts at atmospheric pressure. While the methyl ester yield for the CaO-MgO catalyst with a ratio of 8:2 (CaO:MgO) was 63.6%, it was
E Gershonov et al.
The Journal of organic chemistry, 74(1), 329-338 (2008-12-05)
A facile solvent-free hydrolysis (chemical destruction) of the warfare agents VX (O-ethyl S-2-(diisopropylamino)ethyl methylphosphonothioate), GB (O-isopropyl methylphosphonofluoridate or sarin), and HD (2,2'-dichloroethyl sulfide or sulfur mustard) upon reaction with various solid-supported fluoride reagents is described. These solid reagents include different
Mónica Calvo-Polanco et al.
Physiologia plantarum, 135(1), 51-61 (2009-01-06)
Black spruce (Picea mariana), white spruce (Picea glauca) and jack pine (Pinus banksiana) were inoculated with Suillus tomentosus and subjected to potassium fluoride (1 mM KF and 5 mM KF) in the presence and absence of 60 mM NaCl. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service