Skip to Content
Merck
All Photos(3)

Key Documents

229814

Sigma-Aldrich

Potassium fluoride

≥99.97% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
Beilstein:
3902818
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

Assay

≥99.97% trace metals basis

form

powder and chunks

impurities

≤300.0 ppm Trace Metal Analysis

mp

858 °C (lit.)

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mingming Fan et al.
Bioresource technology, 104, 447-450 (2011-12-16)
Transesterification of soybean oil with methanol was carried out in the presence of CaO-MgO and KF-modified CaO-MgO catalysts at atmospheric pressure. While the methyl ester yield for the CaO-MgO catalyst with a ratio of 8:2 (CaO:MgO) was 63.6%, it was
E Gershonov et al.
The Journal of organic chemistry, 74(1), 329-338 (2008-12-05)
A facile solvent-free hydrolysis (chemical destruction) of the warfare agents VX (O-ethyl S-2-(diisopropylamino)ethyl methylphosphonothioate), GB (O-isopropyl methylphosphonofluoridate or sarin), and HD (2,2'-dichloroethyl sulfide or sulfur mustard) upon reaction with various solid-supported fluoride reagents is described. These solid reagents include different
Mónica Calvo-Polanco et al.
Physiologia plantarum, 135(1), 51-61 (2009-01-06)
Black spruce (Picea mariana), white spruce (Picea glauca) and jack pine (Pinus banksiana) were inoculated with Suillus tomentosus and subjected to potassium fluoride (1 mM KF and 5 mM KF) in the presence and absence of 60 mM NaCl. The
Lirong Wen et al.
Journal of combinatorial chemistry, 11(5), 799-805 (2009-07-28)
A series of new functionalized thiochromeno[2,3-b]pyridine derivatives have been synthesized via a sequence of Knoevenagel condensation, Michael addition, cyclization, and intramolecular nucleophilic substitution, which is first catalyzed by KF/neutral Al(2)O(3) cooperated with PEG 6000 under microwave irradiation. Experimental results indicated
Henry C Astley et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(19), 6200-6205 (2015-04-02)
Many organisms move using traveling waves of body undulation, and most work has focused on single-plane undulations in fluids. Less attention has been paid to multiplane undulations, which are particularly important in terrestrial environments where vertical undulations can regulate substrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service