Skip to Content
Merck
All Photos(5)

Documents

50981

Sigma-Aldrich

Guanidine thiocyanate

BioReagent, for molecular biology, ≥99%

Synonym(s):

Guanidinium rhodanide, Guanidinium thiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
Beilstein:
3563461
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.52

biological source

synthetic

Quality Level

grade

for molecular biology

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

RNA extraction: suitable

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

color

colorless to white,

mp

115-122 °C
120-122 °C (lit.)

solubility

H2O: 4 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5000 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤5000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

room temp

SMILES string

SC#N.NC(N)=N

InChI

1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H

InChI key

ZJYYHGLJYGJLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Guanidine thiocyanate has been used as a component of the stabilization buffer for RNA lysis. It can be used for deactivation of endogenous RNases leading to denaturation of nucleoprotein complexes. It is also suitable for denaturation of macromolecules such as nucleic acids and proteins and for RNA isolation.
Chaotropic agent and strong denaturant; solubilizes cells.

Biochem/physiol Actions

Guanidine thiocyanate (GTC) plays a role in protein denaturation. It is involved in separation of ribosomal ribonucleic acid (rRNA) from ribosomes and denatures ribonuclease (RNase). GTC has been studied in the unfolding and refolding of fluorescent proteins (FP). It can be used along with phenol and chloroform for extraction of RNA.

Principle

Guanidine thiocyanate interferes with the hydrogen bonds and intramolecular interactions of macromolecules. It solubilizes the proteins resulting in disintegration of cellular structures.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Standard methods for molecular research in Apis mellifera
Evans J D, et al.
Journal of Apicultural Research, 52, 1-54 (2013)
Isolation and Purification of RNA
Surzycki S
Basic Techniques in Molecular Biology, 119-144 (2000)
Nasir Ali et al.
BioMed research international, 2017, 9306564-9306564 (2017-08-09)
Nucleic acid extraction (NAE) plays a vital role in molecular biology as the primary step for many downstream applications. Many modifications have been introduced to the original 1869 method. Modern processes are categorized into chemical or mechanical, each with peculiarities
Siun Chee Tan et al.
Journal of biomedicine & biotechnology, 2009, 574398-574398 (2009-12-17)
Extraction of DNA, RNA, and protein is the basic method used in molecular biology. These biomolecules can be isolated from any biological material for subsequent downstream processes, analytical, or preparative purposes. In the past, the process of extraction and purification
Extraction and Purification of Nucleic Acid using CBNP and PCIA Technique
Soni N
PharmaTutor, 4, 20-22 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service