Skip to Content
Merck
All Photos(1)

Key Documents

46415

Supelco

Acrinathrin

PESTANAL®, analytical standard

Synonym(s):

(S)-Cyano(3-phenoxyphenyl)methyl (Z)-(1R,3S)-2,2-dimethyl[2-(2,2,2-trifluoro-1-trifluoromethylethoxycarbonyl)vinyl]-cyclopropanecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21F6NO5
CAS Number:
Molecular Weight:
541.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)[C@@H](\C=C/C(=O)OC(C(F)(F)F)C(F)(F)F)[C@H]1C(=O)O[C@H](C#N)c2cccc(Oc3ccccc3)c2

InChI

1S/C26H21F6NO5/c1-24(2)18(11-12-20(34)38-23(25(27,28)29)26(30,31)32)21(24)22(35)37-19(14-33)15-7-6-10-17(13-15)36-16-8-4-3-5-9-16/h3-13,18-19,21,23H,1-2H3/b12-11-/t18-,19+,21-/m0/s1

InChI key

YLFSVIMMRPNPFK-WEQBUNFVSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

D Cifuentes et al.
Journal of economic entomology, 105(3), 1006-1018 (2012-07-21)
Control of Frankliniella occidentalis (Pergande) is a serious problem for agriculture all over the world because of the limited range of insecticides that are available. Insecticide resistance in F. occidentalis has been reported for all major insecticide groups. Our previous
M S Túri et al.
Acta veterinaria Hungarica, 47(2), 173-179 (1999-05-27)
The Asian bee mite (Varroa jacobsoni Oud.) causes variable damage in Hungarian apiaries due to the different and changing conditions. Plastic and wooden strips impregnated with synthetic pyrethroid-type active ingredients show a high efficacy against the mites. These treatments, however
P Bielza et al.
Journal of economic entomology, 101(2), 499-503 (2008-05-08)
The fitness costs of spinosad and acrinathrin resistance was investigated in the western flower thrips, Frankliniella occidentalis (Pergande) (Thysanoptera: Thripidae). Fitness studies were conducted on susceptible and resistant strains of F. occidentalis. Resistant females were significantly more fecund (number of
Residual fate of acrinathrin in tea under East-Indian climatic condition.
K Saha et al.
Bulletin of environmental contamination and toxicology, 73(4), 713-716 (2004-09-25)
P Bielza et al.
Journal of economic entomology, 102(1), 393-397 (2009-03-04)
The insecticidal efficacy of mixtures of acrinathrin (pyrethroid) with carbamate fungicides (propamocarb, carbendazim, iprovalicarb, and diethofencarb) and insecticides (carbaryl, thiodicarb, pirimicarb, and oxamyl) was studied in a field strain of Frankliniella occidentalis (Pergande). The fungicide propamocarb and the insecticides pirimicarb

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service