Skip to Content
Merck
All Photos(1)

Key Documents

45441

Supelco

Dichlorvos

PESTANAL®, analytical standard

Synonym(s):

2,2-Dichlorovinyl dimethyl phosphate, DDVP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7Cl2O4P
CAS Number:
Molecular Weight:
220.98
Beilstein:
1709141
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

COP(=O)(OC)O\C=C(\Cl)Cl

InChI

1S/C4H7Cl2O4P/c1-8-11(7,9-2)10-3-4(5)6/h3H,1-2H3

InChI key

OEBRKCOSUFCWJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Zhangqian Wang et al.
Scientific reports, 9(1), 411-411 (2019-01-25)
This study was performed to determine the toxicity and possible mechanism of the larvicidal action of honokiol, extracted from Magnolia denudata seeds, and its 10 related compounds against third-instar larvae of insecticide-susceptible Culex pipiens pallens, Aedes aegypti, and Aedes albopictus
Mazarin Akami et al.
Scientific reports, 9(1), 6435-6435 (2019-04-25)
Bacteria inhabiting the gut of insects provide many benefits to their hosts, such as aiding in food digestion, reproduction, and immunity, tissue homeostasis, adaptation to environment and resistance to pathogen and pesticides. The cowpea beetle, Callosobruchus maculatus, is a serious
A S Wright et al.
Archives of toxicology, 42(1), 1-18 (1979-04-23)
The chemical structure, reactivity and metabolic fate of the insecticide dichlorvos (2,2-dichlorovinyl dimethyl phosphate) are discussed in relation to the possible genotoxicity of this and other methyl phosphate triesters. Recent attempts to demonstrate the methylation of DNA following exposure of
Ewan D Booth et al.
Regulatory toxicology and pharmacology : RTP, 49(3), 316-326 (2007-10-16)
Dichlorvos has been in widespread use as an insecticide for over 40 years, during which time its carcinogenicity and genotoxicity have been evaluated extensively. In vitro genotoxicity assays--have shown dichlorvos to be a direct acting genotoxicant at high concentrations, consistent
Nisharg Golash et al.
Ultrasonics sonochemistry, 19(5), 1051-1060 (2012-03-30)
The present work deals with application of sonochemical reactors for the degradation of dichlorvos containing wastewaters. The sonochemical reactor used in the work is a simple ultrasonic horn type operating at 20 kHz with a power rating of 270 W.

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service