Skip to Content
Merck
All Photos(1)

Key Documents

27816

Sigma-Aldrich

Brilliant Blue R

250, for microscopy

Synonym(s):

Acid Blue 83, Brilliant indocyanin 6B, Coomassie Brilliant Blue R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H44N3NaO7S2
CAS Number:
Molecular Weight:
825.97
Colour Index Number:
42660
Beilstein:
5718025
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.26

grade

for microscopy

Quality Level

form

powder

technique(s)

microbe id | staining: suitable

λmax

589.2 nm in ethanol

suitability

suitable for microscopy

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=C/C(C=C5)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C45H45N3O7S2.Na/c1-4-47(31-33-9-7-11-43(29-33)56(49,50)51)40-23-15-36(16-24-40)45(35-13-19-38(20-14-35)46-39-21-27-42(28-22-39)55-6-3)37-17-25-41(26-18-37)48(5-2)32-34-10-8-12-44(30-34)57(52,53)54;/h7-30H,4-6,31-32H2,1-3H3,(H2,49,50,51,52,53,54);/q;+1/p-1

InChI key

NKLPQNGYXWVELD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For detection of protein bands following electrophoresis.

Other Notes

Protein stain for gel electrophoresis; 5 to 10 times more sensitive than Naphthol Blue Black B and Bromophenol (as little as 0.5 μg/cm2 of protein could be detected in a gel matrix)

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camilla Pilotti et al.
Investigative ophthalmology & visual science, 61(5), 19-19 (2020-05-15)
The 32W and 32Q variants of complement factor B (CFB) are associated with reduced risk of developing neovascular age-related macular degeneration (AMD) compared with the common 32R allele. The objective of this study was to determine if the most protective
A procedure for rapid and sensitive staining of protein fractionated by polyacrylamide gel electrophoresis.
A Chrambach et al.
Analytical biochemistry, 20(1), 150-154 (1967-07-01)
Jie Zang et al.
Journal of experimental botany, 71(19), 5896-5910 (2020-07-21)
Iron (Fe) is an essential micronutrient and plays an irreplaceable role in plant growth and development. Although its uptake and translocation are important biological processes, little is known about the molecular mechanism of Fe translocation within seed. Here, we characterized
Virginia Caponera et al.
Journal of economic entomology, 113(1), 348-352 (2019-10-08)
Damage from termite infestations is economically significant and control can be costly when requiring the widespread use of conventional insecticides. Erythritol, a polyalcohol sweetener that is safe for human consumption, causes increased mortality when ingested by some insects, indicating potential
S Fazekas de St Groth et al.
Biochimica et biophysica acta, 71, 377-391 (1963-05-14)
Two new procedures are described for the estimation of protein by direct photometry on electrophoretic strips. The protein complexes of procion brilliant blue RS and coomassie brilliant blue R250 are shown to follow Beer's law up to 50 and 20

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service