Skip to Content
Merck
All Photos(1)

Key Documents

B6529

Sigma-Aldrich

Brilliant Blue R Staining Solution

ethanol solution

Synonym(s):

Brilliant Blue R, Acid Blue 83, Brilliant indocyanin 6B, Coomassie Brilliant Blue R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H44N3NaO7S2
CAS Number:
Molecular Weight:
825.97
Colour Index Number:
42660
Beilstein:
5718025
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Brilliant Blue R Staining Solution, suitable for (for immunoelectrophoresis protein staining)

form

liquid

Quality Level

technique(s)

microbe id | staining: suitable

color

dark blue

suitability

suitable for (for immunoelectrophoresis protein staining)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=C/C(C=C5)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C45H45N3O7S2.Na/c1-4-47(31-33-9-7-11-43(29-33)56(49,50)51)40-23-15-36(16-24-40)45(35-13-19-38(20-14-35)46-39-21-27-42(28-22-39)55-6-3)37-17-25-41(26-18-37)48(5-2)32-34-10-8-12-44(30-34)57(52,53)54;/h7-30H,4-6,31-32H2,1-3H3,(H2,49,50,51,52,53,54);/q;+1/p-1

InChI key

NKLPQNGYXWVELD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Brilliant Blue R staining solution is especially designed for use in staining protein in agarose gels following immunoelectrophoresis and Ouchterlony gels. The stain contains ethanol and acetic acid so gels do not require fixing prior to staining. The immunoelectrophoresis or Ouchterlony gel is first washed with water and saline to remove non-precipitated proteins and then dried. The gel is then immersed in staining solution for 30 min and destained with 10% acetic acid.
For detection of protein bands following electrophoresis.

Components

0.5% (w/v) Brilliant Blue R, 45% (v/v) ethanol and 10% (v/v) acetic acid.

Analysis Note

Tested for suitability on agarose gels following immunoelectrophoresis.

Legal Information

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

81.0 °F - closed cup

Flash Point(C)

27.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zhang et al.
International journal of oncology, 54(5), 1719-1733 (2019-03-14)
Ovarian cancer remains the most lethal type of cancer among all gynecological malignancies. The majority of patients are diagnosed with ovarian cancer at the late stages of the disease. Therefore, there exists an imperative need for the development of early
G Houen et al.
Electrophoresis, 18(5), 701-705 (1997-05-01)
A method for staining proteins on polyvinylidene difluoride membranes without using organic solvent is described. The method uses preblocking of the membrane with either Tween 20 or polyethylene glycol followed by staining with 0.01% Coomassie Brilliant Blue. No destaining of
G Sreeramulu et al.
Electrophoresis, 16(3), 362-365 (1995-03-01)
A novel method for destaining of polyacrylamide gels, stained with Coomassie Brilliant Blue R-250, is described, based on the use of 0.5 M NaCl in water as the destainer, requiring only 2-3 h. Concentrated (> 2 M) or dilute (<
Melda Altikatoglu et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 39(3), 185-190 (2010-12-02)
Horseradish peroxidase (EC 1.11.1.7) was chemically modified by periodate-activated dextran. The activities of free and modified enzyme against organic-aqueous interface and some chemicals were determined. Modified HRP remained fully active in the presence of organic solvent for 4 h. However
Yong-Hak Kim et al.
Archives of biochemistry and biophysics, 494(2), 159-165 (2009-12-01)
We studied the decolorization of malachite green (MG) by the fungus Cunninghamella elegans. The mitochondrial activity for MG reduction was increased with a simultaneous increase of a 9-kDa protein, called CeCyt. The presence of cytochrome c in CeCyt protein was

Articles

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service