Skip to Content
Merck
All Photos(1)

Key Documents

17236

Sigma-Aldrich

3-(Benzyldimethylammonio)propanesulfonate

BioXtra, ≥99.0% (HPCE)

Synonym(s):

3-(N-Phenylmethyl-N,N-dimethylammonio)propanesulfonate, NDSB 256, PDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N+(CH3)2CH2CH2CH2SO3
CAS Number:
Molecular Weight:
257.35
Beilstein:
9194882
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

zwitterionic

Quality Level

product line

BioXtra

Assay

≥99.0% (HPCE)

form

powder

solubility

H2O: 10 mg/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

C[N+](C)(CCCS([O-])(=O)=O)Cc1ccccc1

InChI

1S/C12H19NO3S/c1-13(2,9-6-10-17(14,15)16)11-12-7-4-3-5-8-12/h3-5,7-8H,6,9-11H2,1-2H3

InChI key

MEJASPJNLSQOAG-UHFFFAOYSA-N

Other Notes

Ampholytic detergent used to enhance solubility of proteins in electrophoresis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarita Pudasaini et al.
Frontiers in microbiology, 8, 591-591 (2017-04-26)
Browning Peninsula is an ice-free polar desert situated in the Windmill Islands, Eastern Antarctica. The entire site is described as a barren landscape, comprised of frost boils with soils dominated by microbial life. In this study, we explored the microbial
Tanya R Copley et al.
PloS one, 12(9), e0184095-e0184095 (2017-09-07)
Rhizoctonia solani Kühn infects most plant families and can cause significant agricultural yield losses worldwide; however, plant resistance to this disease is rare and short-lived, and therefore poorly understood, resulting in the use of chemical pesticides for its control. Understanding
Xiaojia Hu et al.
BMC genomics, 18(1), 674-674 (2017-09-02)
The biological control agent Aspergillus aculeatus Asp-4 colonizes and degrades sclerotia of Sclerotinia sclerotiorum resulting in reduced germination and disease caused by this important plant pathogen. Molecular mechanisms of mycoparasites underlying colonization, degradation, and reduction of germination of sclerotia of
Monika Dymarska et al.
PloS one, 12(10), e0184885-e0184885 (2017-10-06)
Entomopathogenic fungi are known for their ability to carry out glycosylation of flavonoids, which usually results in the improvement of their stability and bioavailability. In this study we used a newly isolated strain of the entomopathogenic filamentous fungus Isaria fumosorosea
D Custers et al.
Talanta, 164, 490-502 (2017-01-22)
Erectile dysfunction (ED) is a sexual disorder characterized by the inability to achieve or maintain a sufficiently rigid erection. Despite the availability of non-invasive oral treatment options, many patients turn to herbal alternatives. Furthermore, herbal supplements are increasingly gaining popularity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service