Skip to Content
Merck

Skip To

09963

(Hydroxypropyl)methyl cellulose

Synonym(s):

2-Hydroxypropyl methyl cellulose ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

composition

hydroxylpropoxyl content, ~9%

color

faint beige

viscosity

~15 mPa.s, 2 % in H2O(25 °C)

solubility

water: 100 mg/10 mL, clear, colorless

storage temp.

room temp

SMILES string

O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3COCC(O)C)OCC(O)C)OCC(O)C)OCC(O)C)O[C@@H]4[C@H](O[C@H]([C@@H]([C@H]4OCC(O)C)OCC(O)C)OCC(O)C)COCC(O)C.O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1COC)OC)OC)OC)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2OC)OC)OC)COC

InChI

1S/C36H70O19.C20H38O11/c1-19(37)9-45-17-27-29(47-11-21(3)39)31(48-12-22(4)40)34(51-15-25(7)43)36(54-27)55-30-28(18-46-10-20(2)38)53-35(52-16-26(8)44)33(50-14-24(6)42)32(30)49-13-23(5)41;1-21-9-11-13(23-3)15(24-4)18(27-7)20(30-11)31-14-12(10-22-2)29-19(28-

InChI key

PUSNGFYSTWMJSK-GSZQVNRLSA-N

Application

(Hydroxypropyl)methyl cellulose (HPMC), an inert viscoelastic polymer is used as an emulsifier, thickening and suspending agent similar to animal gelatin. HPMC is used in the development of controlled-delivery medicament.
Thickener for aqueous and non-aqueous systems, clear films with grease resistance, binders, lubricants, steric stabilizer and water retention aid.

Features and Benefits

Dissolves in water, undergoes reversible gelation upon heating, non-ionic, does not complex with ionic species and is surface active and enzyme resistant. Solutions are pseudoplastic.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
423181H7509423173
solubility

water: 100 mg/10 mL, clear, colorless

solubility

-

solubility

water: 10 mg/mL, clear to very slightly hazy, colorless

solubility

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

form

powder

form

powder or crystals

form

powder

form

powder

storage temp.

room temp

storage temp.

-

storage temp.

room temp

storage temp.

-

color

faint beige

color

-

color

white to off-white

color

-

viscosity

~15 mPa.s, 2 % in H2O(25 °C)

viscosity

15,000 cP, 2 wt. % in H2O(20 °C, Ubbelohde)(lit.)

viscosity

2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)

viscosity

100,000 cP, 2 wt. % in H2O(20 °C, Ubbelohde)(lit.)


Still not finding the right product?

Explore all of our products under (Hydroxypropyl)methyl cellulose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
09963-100G04061838658470
09963-25G04061838658487
09963-500G04061838658494

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters