Skip to Content
Merck
All Photos(5)

Documents

S6506

Sigma-Aldrich

Silver nitrate

ReagentPlus®, ≥99.0% (titration)

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor density

5.8 (vs air)

product line

ReagentPlus®

Assay

≥99.0% (titration)

form

solid

mp

212 °C (dec.) (lit.)

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nitrate, also known as Nitric acid silver(I) salt, is an odorless, colorless, or white versatile inorganic compound widely used as an oxidant and cauterizing agent due to its sclerosing, dehydrating, and escharotic properties. It is commonly used as a catalyst in the hydrolytic oxidation of organosilanes, leading to the production of high hydrogen yield and organosilanols.

Application

Silver nitrate can be used as a catalyst:
  • For the oxidation of aromatic, aliphatic, and conjugated aldehydes to the corresponding carboxylic acids in the presence of H2O2 as an oxidant.
  • In the air oxidation of benzylic and allylic alcohols to corresponding aldehydes or ketones in the presence of Na2CO3.
It can also be used as a reagent to synthesize aroyl cyanides from α-bromoarylacetonitriles.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic hydrogen evolution from hydrolytic oxidation of organosilanes with silver nitrate catalyst
Alan Kay Liang T, et al.
Royal Society of Chemistry Advances, 4, 37645-37648 (2014)
Oxidation of aniline with silver nitrate accelerated by p-phenylenediamine: a new route to conducting composites
Patrycja B, et al.
Macromolecules, 43, 10406-10413 (2010)
Revitalising silver nitrate for caries management
Sherry Shiqian G, et al.
International Journal of Environmental Research and Public Health, 15, 80-80 (2018)
Ali Hebeish et al.
Carbohydrate polymers, 92(1), 407-413 (2012-12-12)
Innovative CMC-based hydrogels with great potentials for usage in medical area were principally synthesized as per two strategies .The first involved reaction of epichlorohydrin in alkaline medium containing silver nitrate to yield silver nano-particles (AgNPs)-loaded CMC hydrogel. While CMC acted
Tao Xu et al.
Organic letters, 14(21), 5416-5419 (2012-10-24)
A silver-catalyzed intramolecular oxidative aminofluorination of alkynes has been developed by using NFSI as a fluorinating reagent. This reaction represents an efficient method for the synthesis of various 4-fluoroisoquinolines and 4-fluoropyrrolo[α]isoquinolines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service