Skip to Content
Merck
All Photos(1)

Key Documents

85228

Sigma-Aldrich

Silver nitrate

puriss. p.a., ≥99.5% (AT)

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5% (AT)
form:
solid
Pricing and availability is not currently available.

vapor density

5.8 (vs air)

Quality Level

grade

puriss. p.a.

Assay

≥99.5% (AT)

form

solid

mp

212 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤2 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤1 mg/kg

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nitrate (AgNO3) is commonly used as a precursor to many other silver compounds.

Application

Silver nitrate is used as a precursor in the preparation of:
  • Highly sensitive silver colloids.
  • Chitosan silver nanocomposite (CSN) films.
  • Silver nanoparticles (AgNPs), using sodium borohydride (NaBH4) as a reducing agent and polyethylene glycol (PEG) as stabilizing agent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 988-988 (1994)
A new method for fast preparation of highly surface-enhanced Raman scattering (SERS) active silver colloids at room temperature by reduction of silver nitrate with hydroxylamine hydrochloride.
Leopold N and Lendl B.
The Journal of Physical Chemistry B, 107(24), 5723-5727 (2003)
C Campagnoli et al.
Blood, 98(8), 2396-2402 (2001-10-06)
Human mesenchymal stem/progenitor cells (MSCs) have been identified in adult bone marrow, but little is known about their presence during fetal life. MSCs were isolated and characterized in first-trimester fetal blood, liver, and bone marrow. When 10(6) fetal blood nucleated
D D Songstad et al.
Plant cell reports, 7(4), 262-265 (1988-06-01)
The effect of the ethylene antagonists norbornadiene and silver nitrate and the ethylene precursor l-aminocyclopropane-l-carboxylic acid (ACC) on Zea mays plant regeneration was studied. A 12-fold increase in plant regeneration, as measured by number of plants obtained per gram fresh
Irina Blinova et al.
Environmental science and pollution research international, 20(5), 3456-3463 (2012-11-13)
Although silver nanoparticles (NPs) are increasingly used in various consumer products and produced in industrial scale, information on harmful effects of nanosilver to environmentally relevant organisms is still scarce. This paper studies the adverse effects of silver NPs to two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service