Skip to Content
Merck
All Photos(6)

Key Documents

P3786

Sigma-Aldrich

Potassium phosphate dibasic

Potassium phosphate dibasic
1 of 1 reviewers received a sample product or took part in a promotion

ACS reagent, ≥98%

Synonym(s):

Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38070212
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent

Agency

suitable for SM 5210

Assay

≥98%

form

powder

impurities

≤0.001% Nitrogen compounds (as N)
≤0.01% Insoluble matter

loss

≤1.0% loss on drying, 105°C

pH

8.5-9.6 (25 °C, 5% in solution)

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
Na: ≤0.05%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium phosphate dibasic has been used in the preparation of MMG (maltose, methyl green) medium, phosphate buffered saline and hypotonic solution.[1][2][3]

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and Analysis of Flow-regulated Ion Channels in Xenopus Oocytes.
Shi S
Bio-protocol null
Expression and Purification of the Arabidopsis E4 SUMO Ligases PIAL1 and PIAL2.
Tomanov K and Bachmair A
Bio-protocol null
A new semi-selective medium for Xanthomonas campestris pv. vitians, the causal agent of bacterial leaf spot of lettuce.
Toussaint VCE et al.
Plant Disease, 85, 131-136 (2001)
Sopio Melikishvili et al.
Food chemistry, 337, 127759-127759 (2020-08-11)
High-resolution ultrasonic spectroscopy (HR-US) was applied for real-time monitoring of β-casein hydrolysis by trypsin at various conditions for the first time. The technique is based on the precision measurement of hydration changes proportional to the number of peptide bond hydrolyzed.
Joonhoon Kim et al.
Frontiers in bioengineering and biotechnology, 8, 612832-612832 (2021-02-16)
An oleaginous yeast Rhodosporidium toruloides is a promising host for converting lignocellulosic biomass to bioproducts and biofuels. In this work, we performed multi-omics analysis of lignocellulosic carbon utilization in R. toruloides and reconstructed the genome-scale metabolic network of R. toruloides.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service