Skip to Content
Merck
All Photos(3)

Key Documents

L6883

Sigma-Aldrich

Lithium acetate dihydrate

reagent grade

Synonym(s):

Acetic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOLi · 2H2O
CAS Number:
Molecular Weight:
102.02
Beilstein:
3564320
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
solid

grade

reagent grade

Quality Level

form

solid

composition

Lithium acetate, ≥63%

pH

≤9.5 (5% in solution)

mp

53-56 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤100 ppm

cation traces

Fe: ≤20 ppm
Na: ≤50 ppm

SMILES string

[Li+].[H]O[H].[H]O[H].CC([O-])=O

InChI

1S/C2H4O2.Li.2H2O/c1-2(3)4;;;/h1H3,(H,3,4);;2*1H2/q;+1;;/p-1

InChI key

IAQLJCYTGRMXMA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium acetate dihydrate is one of the commercially available forms of lithium acetate. It is used as a precursor material for the synthesis of various lithium-based compounds. It is also used as a catalyst in several chemical reactions.

Application

Lithium acetate dihydrate (LiOAc.2H2O) can be used as a precursor material to synthesize:      
  • Carbon-modified nanocrystalline LiFePO4 (LiFePO4/C) via sol-gel method using ferrous sulfate, phosphoric acid, citric acid, and polyethylene glycol.      
  • Lithium-graphite nanotubes (LGN) by chemical vapor deposition (CVD) method in the presence of copper with methane as the carbon source.     
  • Conductive polycrystalline solid-state electrolyte (Li4B7O12Cl) by reacting with boric acid and copper(II) chloride dihydrate in trihexyltetradecylphosphonium chloride via an ionothermal method.

It can also be used as a catalyst in the enantioselective synthesis of functionalized 1,3-thiazin-4-ones via N-heterocyclic carbene (NHC)-catalyzed [3 + 3] annulation of thioamides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evelina Tutucci et al.
Nature protocols, 13(10), 2268-2296 (2018-09-16)
The MS2 system has been widely used, in organisms ranging from bacteria to higher eukaryotes, to image single mRNAs in intact cells with high precision. We have recently re-engineered the MS2 system for accurate detection of mRNAs in living Saccharomyces
Zico Alaia Akbar et al.
Optics express, 23(19), A1280-A1287 (2015-09-26)
A unique, hierarchically structured, aggregated TiO(2) nanowire (A-TiO(2)-nw) is prepared by solvothermal synthesis and used as a dual-functioning photoelectrode in dye-sensitized solar cells (DSSCs). The A-TiO(2)-nw shows improved light scattering compared to conventional TiO(2) nanoparticles (TiO(2)-np) and dramatically enhanced dye
Wenyi Hua et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 969, 117-122 (2014-08-30)
A highly sensitive, selective, and rugged quantification method was developed and validated for decitabine (5-aza-2'-deoxycytidine) in human plasma treated with 100μg/mL of tetrahydrouridine (THU). Chromatographic separation was accomplished using hydrophilic interaction liquid chromatography (HILIC) and detection used electrospray ionization (ESI)
Tomasz Krawczyk
Rapid communications in mass spectrometry : RCM, 29(23), 2257-2262 (2015-11-03)
Hexamethylene triperoxide diamine (HMTD) is one of the peroxide-based explosives that are difficult to detect using standard analytical methodologies. It was analyzed by electrospray ionization mass spectrometry (ESI-MS) on a UPLC-TOF instrument. Alkali metal salts were used to promote the
Elaine L Ferguson et al.
Molecular pharmaceutics, 11(12), 4437-4447 (2014-11-02)
Polymer therapeutics offer potential benefits in the treatment of multidrug resistant (MDR) infections; affording targeted delivery of biologically active agents to the site of inflammation, potential decreases in systemic toxicity, and the retention of antimicrobial activity at the target site.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service