Skip to Content
Merck

62393

Lithium acetate dihydrate

BioUltra, ≥99.0% (anhydrous basis)

Synonym(s):

Acetic acid lithium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3COOLi · 2H2O
CAS Number:
Molecular Weight:
102.02
UNSPSC Code:
12352302
NACRES:
NA.26
PubChem Substance ID:
EC Number:
208-914-3
Beilstein/REAXYS Number:
3564320
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Segment

assay

≥99.0% (anhydrous basis)

form

solid

impurities

insoluble matter, passes filter test, 34.0-37.0% water

pH

6.5-8.5 (25 °C, 1 M in H2O)

mp

53-56 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: <0.04, λ: 280 nm Amax: <0.03

SMILES string

[Li+].[H]O[H].[H]O[H].CC([O-])=O

InChI

1S/C2H4O2.Li.2H2O/c1-2(3)4;;;/h1H3,(H,3,4);;2*1H2/q;+1;;/p-1

InChI key

IAQLJCYTGRMXMA-UHFFFAOYSA-M

Other Notes

In the preparation of the modified ninhydrin reagent for amino analysis by ion-exchange chromatography; Essential for obtaining a fully soluble dimethyl sulfoxide-buffer mixture and for eliminating possible salt precipitation when the ninhydrin reagent mixes with the sodium citrate-containing effluent from an ion-exchange column.


Still not finding the right product?

Explore all of our products under Lithium acetate dihydrate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library