Skip to Content
Merck
All Photos(1)

Key Documents

95682

Sigma-Aldrich

p-Xylene

puriss. p.a., ≥99.0% (GC)

Synonym(s):

1,4-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1901563
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

vapor density

3.7 (vs air)

vapor pressure

9 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

984 °F

expl. lim.

7 %

impurities

≤0.5% o- and m-xylene (GC)
≤0.5% toluene (GC)

evapn. residue

≤0.002%

refractive index

n20/D 1.495 (lit.)
n20/D 1.495

bp

138 °C (lit.)

mp

12-13 °C (lit.)

solubility

water: soluble 0.2 g/L

density

0.861 g/mL at 20 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

Cc1ccc(C)cc1

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3

InChI key

URLKBWYHVLBVBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Xylene is an isomeric form of xylene. It undergoes Friedel-Crafts acylation reaction with various acids in the presence of zeolites (catalyst). It can be obtained from the mixtures of isomeric forms of xylene and ethylbenzene, isolated from the naptha fraction of petroleum. Also, it can be prepared from toluene by employing ZSM (Zeolite Socony Mobil)-5 catalysts. It is a raw material for the manufacture of PET (poly(ethylene terephthalate)) bottles. Sustainable synthesis of xylene from ethylene and dimethylfuran in the presence of zeolite catalysts has been reported.

Application

p-Xylene may be used in the synthesis of poly-p-xylene and terephthalic acid.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra-selective cycloaddition of dimethylfuran for renewable p-xylene with H-BEA.
Chang CC, et al.
Green Chemistry, 16(2), 585-588 (2014)
Cycloaddition of biomass-derived furans for catalytic production of renewable p-xylene.
Williams CL, et al.
ACS Catalysis, 2(6), 935-939 (2012)
Friedel-Crafts acylation of toluene and p-xylene with carboxylic acids catalyzed by zeolites.
Chiche B, et al.
The Journal of Organic Chemistry, 51(11), 2128-2130 (1986)
Metal-organic framework MIL-101 for high-resolution gas-chromatographic separation of xylene isomers and ethylbenzene.
Zhi-Yuan Gu et al.
Angewandte Chemie (International ed. in English), 49(8), 1477-1480 (2010-01-22)
Sergio Grunder et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(49), 15632-15649 (2012-10-24)
Molecular gauge blocks, based on 1-7, 9-11 paraxylene rings, have been synthesized as part of a homologous series of oligoparaxylenes (OPXs) with a view to providing a molecular tool box for the construction of nano architectures-such as spheres, cages, capsules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service