Skip to Content
Merck
All Photos(2)

Key Documents

247642

Sigma-Aldrich

Xylenes

ACS reagent, ≥98.5% xylenes + ethylbenzene basis

Synonym(s):

Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
137-140 °C (lit.)
vapor pressure:
18 mmHg ( 37.7 °C)

grade

ACS reagent

Quality Level

vapor density

3.7 (vs air)

vapor pressure

18 mmHg ( 37.7 °C)

Assay

≥98.5% xylenes + ethylbenzene basis

form

liquid

composition

C6H5CH2CH3, ≤25%

expl. lim.

7 %

dilution

(for analytical testing)

impurities

H2SO4, passes test
≤0.003% S compounds
≤0.05% water

evapn. residue

≤0.002%

color

APHA: ≤10

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

mp

-34 °C

density

0.86 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xylenes is a mixture of isomeric ortho-,meta- and para- forms of xylene which is an aromatic hydrocarbon widelyused as a solvent in various chemical reactions.

Application

Xylenes can be used as a solvent:
  • In the tetraphosphine/palladium-catalyzed Suzuki cross-coupling with sterically hindered aryl halides and arylboronic acids to synthesize tri-ortho-substituted biaryl adducts.
  • For the N-alkylation of amines or ammonia with alcohols to synthesize N-alkyl amines.
  • To synthesize aromatic polyimides by reacting unsymmetric phosphinated diamines and dianhydrides via solution polymerization.

Packaging

poly-coated bottles available: product numbers 247642-1L-CB, 247642-4L-CB and 247642-4X4L-CB

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A study of nonrigid aromatic molecules by supersonic molecular jet spectroscopy. I. Toluene and the xylenes.
Breen PJ, et al.
J. Chem. Phys. , 87(4), 1917-1926 (1987)
Transport and isomerization of xylenes over HZSM-5 zeolites.
Mirth G, et al.
J. Catal., 139(1), 24-33 (1993)
Magdalena Pasarica et al.
Diabetes, 58(3), 718-725 (2008-12-17)
Based on rodent studies, we examined the hypothesis that increased adipose tissue (AT) mass in obesity without an adequate support of vascularization might lead to hypoxia, macrophage infiltration, and inflammation. Oxygen partial pressure (AT pO2) and AT temperature in abdominal
Qiming Wang et al.
Nature communications, 5, 4899-4899 (2014-09-17)
Cephalopods can display dazzling patterns of colours by selectively contracting muscles to reversibly activate chromatophores--pigment-containing cells under their skins. Inspired by this novel colouring strategy found in nature, we design an electro-mechano-chemically responsive elastomer system that can exhibit a wide
Zifu Li et al.
Chemical communications (Cambridge, England), 47(1), 331-333 (2010-08-24)
We describe a flexible method for large-scale production of polymeric microspheres by templating a microgel-stabilized oil-in-water (o/w) high internal phase emulsion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service