Skip to Content
Merck
All Photos(1)

Documents

31807

Sigma-Aldrich

Cesium chloride

puriss. p.a., ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

Assay

≥99.5%

form

crystals

impurities

≤0.001% total nitrogen (N)

pH

6-7.5

mp

645 °C (lit.)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ba: ≤20 mg/kg
Ca: ≤20 mg/kg
Cu: ≤2 mg/kg
Fe: ≤5 mg/kg
K: ≤20 mg/kg
Li: ≤0.5 mg/kg
Mg: ≤5 mg/kg
Na: ≤20 mg/kg
Pb: ≤1 mg/kg
Rb: ≤50 mg/kg
Zn: ≤2 mg/kg

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cesium chloride is a cesium halide. Cesium halides can be prepared by reacting cesium carbonate with corresponding hydrohalic acids. Cesium chloride ultracentrifugation method has been reported for the extraction of RNA from cellular fractions.

Application

Cesium chloride may be employed in gel-electrophoresis† and RNA purification studies.† It may be employed for the isolation of bacterial plasmid from Agrobacterium spp.†
Used for the preparation of electrically conducting glasses.Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ribonucleic acid isolated by cesium chloride centrifugation.
Glisin V, et al.
Biochemistry, 13(12), 2633-2637 (1974)
Therese Abrahamsson et al.
Neuron, 73(6), 1159-1172 (2012-03-27)
Interneurons are critical for neuronal circuit function, but how their dendritic morphologies and membrane properties influence information flow within neuronal circuits is largely unknown. We studied the spatiotemporal profile of synaptic integration and short-term plasticity in dendrites of mature cerebellar
Juan Armendáriz-Borunda et al.
Journal of bioscience and bioengineering, 112(5), 415-421 (2011-08-23)
Gene therapy represents a promising approach in the treatment of several diseases. Currently, the ideal vector has yet to be designed; though, adenoviral vectors (Ad-v) have provided the most utilized tool for gene transfer due principally to their simple production
Sandra Aedo et al.
Antimicrobial agents and chemotherapy, 56(11), 5458-5464 (2012-08-08)
DNA topoisomerases are important targets in anticancer and antibacterial therapy because drugs can initiate cell death by stabilizing the transient covalent topoisomerase-DNA complex. In this study, we employed a method that uses CsCl density gradient centrifugation to separate unbound from
Shengyuan Ding et al.
Journal of neurophysiology, 106(6), 3019-3034 (2011-09-02)
GABA projection neurons (GABA neurons) in the substantia nigra pars reticulata (SNr) and dopamine projection neurons (DA neurons) in substantia nigra pars compacta (SNc) have strikingly different firing properties. SNc DA neurons fire low-frequency, long-duration spikes, whereas SNr GABA neurons

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service