Skip to Content
Merck
All Photos(4)

Key Documents

C3309

Sigma-Aldrich

Cesium chloride

BioXtra, ≥99.5% (titration)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

product line

BioXtra

Assay

≥99.5% (titration)

form

powder

impurities

Insoluble matter, passes filter test

loss

≤0.5% loss on drying, 110 °C

pH

5.0-7.5 (20 °C, 3 M in H2O)

mp

645 °C (lit.)

solubility

H2O: 3 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
As: ≤0.00001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.005%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.02%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.03 at 260 in H2O at 3 M
≤0.03 at 280 in H2O at 3 M

application(s)

hematology
histology

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used for the preparation of electrically conducting glasses.Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Jurkiewicz et al.
Biochimica et biophysica acta, 1818(3), 609-616 (2011-12-14)
Effects of alkali metal chlorides on the properties of mixed negatively charged lipid bilayers are experimentally measured and numerically simulated. Addition of 20mol% of negatively charged phosphatidylserine to zwitterionic phosphatidylcholine strengthens adsorption of monovalent cations revealing their specificity, in the
Takafumi Yamamoto et al.
Inorganic chemistry, 50(22), 11787-11794 (2011-10-25)
The rock salt (B1) structure of binary oxides or chalcogenides transforms to the CsCl (B2) structure under high pressure, with critical pressures P(s) depending on the cation to anion size ratio (R(c)/R(a)). We investigated structural changes of A(2)MO(3) (A =
Sandra Aedo et al.
Antimicrobial agents and chemotherapy, 56(11), 5458-5464 (2012-08-08)
DNA topoisomerases are important targets in anticancer and antibacterial therapy because drugs can initiate cell death by stabilizing the transient covalent topoisomerase-DNA complex. In this study, we employed a method that uses CsCl density gradient centrifugation to separate unbound from
Lei Li et al.
Cell reports, 28(11), 2979-2995 (2019-09-12)
Munc13 proteins play several roles in regulating short-term synaptic plasticity. However, the underlying molecular mechanisms remain largely unclear. Here we report that C. elegans UNC-13L, a Munc13-1 ortholog, has three domains that inhibit synaptic vesicle (SV) exocytosis. These include the X
Philip A Gurnev et al.
Langmuir : the ACS journal of surfaces and colloids, 28(45), 15824-15830 (2012-10-24)
We demonstrate that the cation-selective channel formed by gramicidin A can be used as a reliable sensor for studying the multivalent ion accumulation at the surfaces of charged lipid membranes and the "charge inversion" phenomenon. In asymmetrically charged membranes with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service