Skip to Content
Merck
All Photos(3)

Key Documents

31445

Sigma-Aldrich

Sodium peroxide

puriss. p.a., ACS reagent, reag. ISO, ≥95%

Synonym(s):

Sodium superoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O2
CAS Number:
Molecular Weight:
77.98
EC Number:
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥95%
form:
powder

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. ISO

Assay

≥95%

form

powder

impurities

≤0.002% heavy metals (as Pb)
≤0.003% total nitrogen (N)

pH

12.8 (20 °C, 100 g/L)

mp

460 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤10 mg/kg
Fe: ≤20 mg/kg

SMILES string

[Na+].[Na+].[O-][O-]

InChI

1S/2Na.O2/c;;1-2/q2*+1;-2

InChI key

PFUVRDFDKPNGAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium peroxide (Sodium superoxide) has been synthesized by the matrix reaction of sodium atoms and oxygen molecules at high dilution in argon. Its IR spectral studies have been reported.

Application

Sodium peroxide may be used for the isotope dilution inductively coupled plasma mass spectrometric quantification of Ru, Pd, Ir, Pt and rare earth elements (such as Y, Th, Zr, Hf, Nb and Ta) in various geological samples.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of platinum, palladium, ruthenium and iridium in geological samples by isotope dilution inductively coupled plasma mass spectrometry using a sodium peroxide fusion and tellurium coprecipitation
Enzweiler J, et al.
Analyst, 120(5), 1391-1396 (1995)
Infrared spectra and bonding in the sodium superoxide and sodium peroxide molecules.
Andrews L.
The Journal of Physical Chemistry, 73(11), 3922-3928 (1969)
Determination of Rare Earth Elements, Y, Th, Zr, Hf, Nb and Ta in Geological Reference Materials G-2, G-3, SCo-1 and WGB-1 by Sodium Peroxide Sintering and Inductively Coupled Plasma-Mass Spectrometry.
Meisel T, et al.
Geostandards Newsletter, 26(1), 53-61 (2002)
R J Calvert et al.
The American journal of clinical nutrition, 49(5), 901-903 (1989-05-01)
A safer method of oxidation of Cr2O3-containing fecal samples from transit-time studies was developed using sodium peroxide to replace perchloric acid as the oxidizing agent. The percentage recovery of Cr2O3 with this method was compared with that of perchloric acid
K J Sreeram et al.
Journal of environmental monitoring : JEM, 3(5), 526-530 (2001-11-07)
The processing of chromite ore is associated with the generation of large quantities of solid wastes containing chromium, which have been disposed of as landfill for many years. The mobilization and operational speciation of chromium contained in soils contaminated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service