Skip to Content
Merck
All Photos(4)

Key Documents

31232

Sigma-Aldrich

Iron(III) chloride hexahydrate

puriss. p.a., reag. Ph. Eur., ≥99%

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
form:
solid

Agency

USP/NF
reag. Ph. Eur.

Quality Level

vapor pressure

1 mmHg ( 194 °C)

grade

puriss. p.a.

Assay

≥99%

form

solid

impurities

≤0.001% free chlorine (Cl)
≤0.001% total nitrogen (N)
≤0.05% sub.not precip.b.NH4OH(SO4)
≤0.2% free acid (as HCl)

bp

280-285 °C (lit.)

mp

37 °C (lit.)

anion traces

phosphate (PO43-): ≤100 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤5 mg/kg
Cu: ≤20 mg/kg
Fe2+: ≤50 mg/kg
Pb: ≤20 mg/kg
Zn: ≤20 mg/kg

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Iron(III) chloride hexahydrate can be obtained from crystallization of iron(III) chloride from water. Its crystal structure has been studied using three-dimensional single-crystal X-ray-diffraction data. Its crystals have two chloride ions and four water molecules arranged around each ferric ion to form octahedral trans-[FeCl2(OH2)4]+ ions having C2h-2/m symmetry.

Application

Iron(III) chloride hexahydrate may be employed as an oxidant for the following conversions:
  • Synthesis of 2-substituted 4(3H)-quinazolinones (yield = 77-93%), via condensation of aldehydes with anthranilamide.
  • Oxidation of 1,4-dihydropyridines (1,4-DHPs) to corresponding pyridines.
  • Oxidation of 4-isopropyl-1,4-DHP to dealkylated pyridine.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferric chloride hexahydrate: a convenient reagent for the oxidation of hantzsch 1, 4-dihydropyridines.
Lu J, et al.
Synthetic Communications, 31(17), 2625-2630 (2001)
Benign and efficient synthesis of 2-substituted 4(3H)-quinazolinones mediated by iron (III) chloride hexahydrate in refluxing water.
Wang G-W, et al.
Bulletin of the Chemical Society of Japan, 79(9), 1426-1430 (2006)
Ievgen V Pylypchuk et al.
Nanomaterials (Basel, Switzerland), 10(2) (2020-02-13)
Sustainable and green synthesis of nanocomposites for degradation of pharmaceuticals was developed via immobilization and stabilization of the biological strong oxidizing agents, peroxidase enzymes, on a solid support. Sol-gel encapsulated enzyme composites were characterized using electron microscopy (TEM, SEM), atomic
Crystal structure of ferric chloride hexahydrate.
Lind MD.
J. Chem. Phys. , 47(3), 990-993 (1967)
Chenglong Yuan et al.
International journal of molecular sciences, 20(3) (2019-02-13)
The prion protein (PrPSc) has drawn widespread attention due to its pathological potential to cause prion diseases. Herein, we successfully synthesized Fe₃C@C by carbonizing Fe₃O₄-lignin clusters, which were prepared through a facile hydrogen bonding interaction between ≡Fe-OH and hydroxyl groups

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service