Skip to Content
Merck
All Photos(1)

Key Documents

80353

Millipore

TDA Reagent

suitable for microbiology

Synonym(s):

Iron(III) chloride solution, Tryptophan Deaminase Reagent, Ferric chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20
MDL number:
UNSPSC Code:
41171621
PubChem Substance ID:
NACRES:
NA.85

Agency

according to ISO 10273:2017

Quality Level

product line

BioChemika

form

liquid

shelf life

limited shelf life, expiry date on the label

composition

ferric chloride, 1 g/10 mL (in distilled water)

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
food and beverages

microbiology

suitability

Proteus spp.

SMILES string

Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe/h3*1H;/q;;;+3/p-3

InChI key

RBTARNINKXHZNM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

For identification of Proteus species by detection of tryptophan deaminase activity.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdelwareth A O Sarhan et al.
Chemical Society reviews, 38(9), 2730-2744 (2009-08-20)
In this critical review, the use of iron(III) chloride in oxidative C-C couplings of arenes and related unsaturated compounds is presented and reviewed. The approach allows highly selective dimerisations of phenol derivatives, naphthols, and heterocyclic compounds. Sequential couplings give access
Chen-Yu Chen et al.
Clinica chimica acta; international journal of clinical chemistry, 438, 337-341 (2014-10-05)
Insulin-like growth factor binding protein-1 (IGFBP-1) constitutes a subgroup of the insulin-like growth factor binding protein systems, and its concentration in amniotic fluid is 100-1000 times higher than the concentration in other body fluids. The aim of this study was
Xiaoqing Li et al.
The Journal of organic chemistry, 78(18), 9499-9504 (2013-08-28)
Halosulfonylation of terminal alkynes was achieved with sulfonylhydrazides as the sulfonyl precursor and inexpensive iron halide as halide source in the presence of TBHP, allowing the regio- and stereoselective generation of (E)-β-chloro and bromo vinylsulfones.
M Golder et al.
Thrombosis and haemostasis, 109(1), 53-60 (2012-11-28)
Factor VIII (FVIII), a procoagulant cofactor, plays a crucial role in the intrinsic coagulation cascade. A causal association between elevated FVIII levels and venous thrombosis incidence has been established; no such association has been confirmed with arterial thrombosis. The independent
Dattatraya H Dethe et al.
Organic letters, 15(3), 429-431 (2013-01-16)
A novel approach was developed for the synthesis of highly substituted indene derivatives, using an FeCl(3) catalyzed Prins-type cyclization reaction which was further applied in the total synthesis of jungianol and epi-jungianol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service