Skip to Content
Merck
All Photos(4)

Key Documents

221783

Sigma-Aldrich

Copper(II) chloride dihydrate

reagent grade

Synonym(s):

Cupric chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2 · 2H2O
CAS Number:
Molecular Weight:
170.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
crystals
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

>1 (vs air)

form

crystals

concentration

35.0-38.0% Cu (EDTA titration)

pH

3-3.8

mp

100 °C (dec.) (lit.)

SMILES string

Cl[Cu]Cl.[H]O[H].[H]O[H]

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Copper(II) chloride dihydrate may be used in the following processes:
  • As a recoverable promoter for the conversion of oximes to carbonyl compounds under mild reaction conditions.[1]
  • To catalyze Biginelli condensation of aldehyde, β-ketoester and urea or thiourea to form dihydropyrimidinones under solvent-free and microwave irradiations.[2]
  • Along with stannous chloride for the allylation of aldehydes.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel Cu-catalysed three-component one-pot synthesis of dihydropyrimidin-2 (1H)-ones using microwaves under solvent-free conditions
Gohain M, et al
Synlett, 2004(02), 0235-0238 (2004)
A green chemistry method for the regeneration of carbonyl compounds from oximes by using cupric chloride dihydrate as a recoverable promoter for hydrolysis
Quan N, et al
Synlett, 2011(07), 1028-1032 (2011)
A novel copper (II)/tin (II) reagent for aqueous carbonyl allylation: In situ diagnostics of reactive organometallics in water
Kundu A, et al
Organometallics, 16(22), 4796-4799 (1997)
Deprotection of t-butyldimethylsiloxy (TBDMS) protecting group with catalytic copper (II) chloride dihydrate.
TAN ZP, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 11(9), 753-756 (2000)
An X-ray refinement of the crystal structure of copper (II) chloride dihydrate.
Engberg ?.
Acta Chemica Scandinavica, 24(10), 6-6 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service