Skip to Content
Merck
All Photos(3)

Key Documents

31307

Sigma-Aldrich

Calcium chloride dihydrate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99%
form:
crystalline
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

0.01 hPa ( 20 °C)

Assay

≥99%

form

crystalline

impurities

≤0.003% oxidizing substances (as NO3)
≤0.005% ammonium (NH4)
≤0.01% insoluble in water
≤5 ppm heavy metals (by ICP)

pH

4.5-8.0 (25 °C, 5%)

mp

176 °C (dec.) (lit.)

anion traces

nitrate (NO3-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤1 mg/kg
Ba: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Sr: ≤100 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium chloride dihydrate is a useful biochemical reagent.

Application

Calcium chloride dihydrate was used in the preparation of calcium-alginate beads.[1] It may be used for the concentration of virus vectors in a protocol.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concentration of viral vectors by co-precipitation with calcium phosphate.
Pham L, et al.
The journal of gene medicine, 3(2), 188-194 (2001)
George Pasparakis et al.
International journal of pharmaceutics, 323(1-2), 34-42 (2006-07-11)
The aim of the present work was to investigate the swelling behavior and the in vitro release of the antihypertensive drug verapamil hydrochloride from calcium alginate and chitosan treated calcium alginate beads. Calcium-alginate beads, chitosan-coated alginate beads and alginate-chitosan mixed
João Barroso-Batista et al.
Current biology : CB, 30(6), 1049-1062 (2020-03-07)
Members of the gut microbiota are thought to experience strong competition for nutrients. However, how such competition shapes their evolutionary dynamics and depends on intra- and interspecies interactions is poorly understood. Here, we test the hypothesis that Escherichia coli evolution
P Geetha et al.
Ecotoxicology and environmental safety, 122, 17-23 (2015-07-15)
Population explosion, depletion of water resources and prolonged droughts and floods due to climatic change lead to scarcity of pure and hygienic drinking water in most of the developing countries. Recently nanomaterials attained considerable attention as biosorbent for water purification
Marianne T Vestermark et al.
Acta orthopaedica, 82(5), 614-621 (2011-09-08)
Allografts are often used during revision hip replacement surgery for stabilization of the implant. Resorption of the allograft may exceed new bone formation, and instability of the prosthesis can develop. We investigated whether strontium could regulate the imbalance of fast

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service