Skip to Content
Merck
All Photos(3)

Key Documents

212865

Sigma-Aldrich

Copper(I) bromide

98%

Synonym(s):

Cuprous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuBr
CAS Number:
Molecular Weight:
143.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
98%
form:
solid
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

reaction suitability

reaction type: click chemistry
reagent type: catalyst
core: copper

mp

504 °C (lit.)

density

4.71 g/mL at 25 °C (lit.)

SMILES string

[Cu]Br

InChI

1S/BrH.Cu/h1H;/q;+1/p-1

InChI key

NKNDPYCGAZPOFS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Copper(I) bromide (CuBr) may be used in the synthesis of the following:
  • enantioenriched, functionalized and protected propargylamines[1]
  • polyvinylchloride bearing benzoxazine side groups[2]
  • polybutadiene having benzoxazine side-groups (PB-benzoxazine)[3]
  • mononuclear complexes [Cu(L)(PPh3)2Br][4]
  • Copper(I) bromide along with N-(n-octyl)-2-pyridylmethanimine participates in the living-radical polymerization of methyl methacrylate.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective synthesis of propargylamines by copper-catalyzed addition of alkynes to enamines.
Christopher Koradin et al.
Angewandte Chemie (International ed. in English), 41(14), 2535-2538 (2002-08-31)
Copper (I) bromide/N-(n-octyl)-2-pyridylmethanimine-mediated living-radical polymerization of methyl methacrylate using carbosilane dendritic initiators.
Hovestad NJ, et al.
Macromolecules, 33(11), 4048-4052 (2000)
Thermally curable polyvinylchloride via click chemistry.
Kiskan B, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 46(11), 3512-3518 (2008)
Copper (I) bromide complexes with heterocyclic thiones and triphenylphosphine as ligands. The X-ray crystal structure of copper (I) pyrimidine-2-thione bis (triphenylphosphine) bromide [Cu (PPh 3) 2 (PymtH) Br].
Lecomte C, et al.
Polyhedron, 8(8), 1103-1109 (1989)
B Podobnik et al.
Bioconjugate chemistry, 26(3), 452-459 (2015-01-30)
The covalent attachment of poly(ethylene glycol) (PEG) to therapeutic proteins is a commonly used approach for extending in vivo half-lives. A potential limitation of this PEGylation strategy is the adverse effect of PEG on conjugate viscosity. Interferon-alpha (IFN) was conjugated

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service