Skip to Content
Merck
All Photos(1)

Key Documents

923060

Sigma-Aldrich

Copper(I) bromide

99.9% trace metals basis

Synonym(s):

Cuprous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuBr
CAS Number:
Molecular Weight:
143.45
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Assay:
99.9% trace metals basis
form:
powder
Pricing and availability is not currently available.

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: copper

mp

504 °C (lit.)

density

4.71 g/mL at 25 °C (lit.)

SMILES string

[Cu]Br

InChI

1S/BrH.Cu/h1H;/q;+1/p-1

InChI key

NKNDPYCGAZPOFS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Copper bromide (CuBr) is used in Sandmeyer’s reaction to prepare aryl bromides from aryl diazonium salts. [1] It is also used as a co-catalyst in Sonogashira coupling reaction. [2] CuBr reacts with organolithium compounds to form organocopper complexes that are commonly used in organic synthesis for C-C bond formation. [3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances and perspectives in copper-catalyzed Sonogashira coupling reactions.
Thomas, A. et al
Royal Society of Chemistry Advances, 4(42), 21688-21698 (2014)
Unravelling the hidden link of lithium halides and application in the synthesis of organocuprates
Yi, H., et al
Nature Communications, 8, 14794 -14794 (2017)
Cu (I)/Cu (II)/TMEDA, new effective available catalyst of sandmeyer reaction
Sigeev, A. S., et al
European Journal of Organic Chemistry, 48(8), 1055-1058 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service