Skip to Content
Merck
All Photos(1)

Key Documents

1.04616

Supelco

Hydroxylammonium chloride

GR for analysis ACS,ISO,Reag. Ph Eur

Synonym(s):

Hydroxylammonium chloride, Hydroxylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H3NO · xHCl
CAS Number:
Molecular Weight:
33.03 (free base basis)
MDL number:
UNSPSC Code:
12352301
EC Index Number:
226-798-2
NACRES:
NB.21
grade:
ACS reagent
vapor pressure:
0.054 Pa ( 50 °C)

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

0.054 Pa ( 50 °C)

Assay

≥99% (manganometric)

form

solid

dilution

(for analytical testing)

pH

2.5-3.5 (20 °C, 50 g/L in H2O)

mp

154 °C

density

1.70 g/cm3 at 20.2 °C

bulk density

900 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].NO.[H+]

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2

InChI key

WTDHULULXKLSOZ-UHFFFAOYSA-N

Analysis Note

Assay (manganometric): ≥ 99.0 %
Clarity of the solution in Ethanol: passes test
pH-value (5 %; water): 2.5 - 3.5
Titrable free acid: ≤ 0.25 meq/g
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (ACS): ≤ 0.0005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
NH₄ (Ammonium): passes test
Pb (Lead): ≤ 0.0005 %
Total sulfur (as sulfate): ≤ 0.005 %
Residue on ignition (as sulfate): ≤ 0.01 %
Corresponds to ACS, ISO, Reag. Ph Eur

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

spleen

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service