Skip to Content
Merck
All Photos(1)

Key Documents

05211

Sigma-Aldrich

Sodium hydroxide solution

purum, ≥32%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

3 mmHg ( 37 °C)

grade

purum

form

solution

concentration

≥32%

technique(s)

titration: suitable

impurities

≤0.001% heavy metals (as Pb)

pH

14

density

1.327 g/cm3 at 25 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Fe: ≤20 mg/kg

SMILES string

[OH-].[Na+]
[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide participates in the oxidation of glycerol catalyzed by Au/charcoal or Au/graphite.[1] It also participates in solvent-free aldol condensation reaction.[2]

Application

Sodium hydroxide may be employed for the determination of specific surface area of colloidal silica, via titration.[3] It may be employed for the direct oxidation of sodium borohydride, which was investigated by cyclic and linear voltammetry, chronoamperometry and chronopotentiometry.[4] It may be used to compose citrate buffer and borate buffers.[5] It may be used in the synthesis of :
  • alkylaryl amidosulfobetaines[6]
  • (3-[4-teroctylphenoxyethoxyethoxyethyl] dimethylammonio propane sulfonate (XOSB)[6]
  • trisulfobetaines (TriSBn)[6]

Other Notes

The article number 05211-4X2.5L will be discontinued. Please order the single bottle 05211-2.5L which is physically identical with the same exact specifications.
The article number 05211-4X5L will be discontinued. Please order the single bottle 05211-5L which is physically identical with the same exact specifications.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of sodium borohydride direct oxidation and oxygen reduction in sodium hydroxide electrolyte: Part I. BH4- electro-oxidation on Au and Ag catalysts.
Chatenet M, et al.
Electrochimica Acta, 51(25), 5459-5467 (2006)
Silvio Carrettin et al.
Chemical communications (Cambridge, England), (7)(7), 696-697 (2002-07-18)
Glycerol is oxidised to glyceric acid with 100% selectivity using either 1% Au/charcoal or 1% Au/graphite catalyst under mild reaction conditions (60 degrees C, 3 h, water as solvent).
Determination of specific surface area of colloidal silica by titration with sodium hydroxide.
Sears Jr GW.
Analytical Chemistry, 28(12), 1981-1983 (1956)
Chemoselective, solvent-free aldol condensation reaction.
Raston CL and Scott JL.
Green Chemistry, 2(2), 49-52 (2000)
M A Roberts et al.
Analytical chemistry, 69(11), 2035-2042 (1997-06-01)
This report describes a UV laser photoablation method for the production of miniaturized liquid-handling systems on polymer substrate chips. The fabrication of fluid channel and reservoir networks is accomplished by firing 200 mJ pulses from an UV excimer laser at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service