Skip to Content
Merck
All Photos(1)

Key Documents

Y0001980

Folic acid for impurity I identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Folic acid, PteGlu, Pteroyl-L-glutamic acid, Vitamin M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N7O6
CAS Number:
Molecular Weight:
441.40
Beilstein:
100781
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

folic acid

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(N1)=NC(C2=C1N=CC(CNC3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=N2)=O

InChI

1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1

InChI key

OVBPIULPVIDEAO-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Folic acid, a water-soluble B vitamin, is found in several food products. As it is not generated naturally in the human body, diet is one of the easiest ways to obtain it. It plays a crucial role in the biochemical synthesis and repair of DNA and is used in the management of megaloblastic anemias, neural tube defects such as anencephaly and spina bifida. Pharmaceutical drug forms of folic acid are desirable for supplementing its consumption through the diet.[1][2]

Application

Folic acid for impurity I identification may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography.[3]

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Flow injection chemiluminometric determination of folic acid in pharmaceutical formulations
Al-Warthan.AA
Analytical Sciences, 10(6), 919-922 (1994)
A UV spectrophotometric method for the determination of folic acid in pharmaceutical tablets and dissolution tests
Matias R, et al.
Analytical Methods : Advancing Methods and Applications, 6(9), 3065-3071 (2014)
Folic acid hydrate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 6(9), 5662-5663 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service