Skip to Content
Merck
All Photos(1)

Documents

F0300000

Folic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

PteGlu, Pteroyl-L-glutamic acid, Vitamin M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N7O6
CAS Number:
Molecular Weight:
441.40
Beilstein:
100781
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

API family

folic acid

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

>285 °C

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(N1)=NC(C2=C1N=CC(CNC3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=N2)=O

InChI

1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1

InChI key

OVBPIULPVIDEAO-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Folic acid, a water-soluble B vitamin, is available in several food products. As it is not generated in the human body, diet is one of the easiest ways to obtain it. It plays a crucial role in the biochemical synthesis and repair of DNA and is used in the management of megaloblastic anemias, neural tube defects such as anencephaly and spina bifida. Pharmaceutical drug forms of folic acid are desirable for supplementing its consumption through the diet.

Application

Folic acid may be used as an European Pharmacopoeia reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Folic acid hydrate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 6(5-6), 5662-5663 (2018)
Flow injection chemiluminometric determination of folic acid in pharmaceutical formulations
Al-Warthan.AA
Analytical Sciences, 10(6), 919-922 (1994)
Adsorptive stripping voltammetric determination of folic acid in pharmaceutical preparations
Szczepaniak W and Ren M
Electroanalysis, 6(5-6), 505-507 (1994)
A UV spectrophotometric method for the determination of folic acid in pharmaceutical tablets and dissolution tests
Matias R, et al.
Analytical Methods : Advancing Methods and Applications, 6(9), 3065-3071 (2014)
Determination of folic acid in tablets by microemulsion electrokinetic chromatography
Aurora-Prado SM, et al.
Journal of Chromatography A, 1051(1-2), 291-296 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service