Skip to Content
Merck
All Photos(1)

Key Documents

Y0001337

Oseltamivir phosphate (impurity B free)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Oseltamivir phosphate, (3R,4R,5S)-4-Acetamido-5-amino-3-(1-ethylpropoxy)-1-cyclohexene-1-carboxylic acid ethyl ester phosphate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28N2O4 ·H3PO4
CAS Number:
Molecular Weight:
410.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oseltamivir

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OP(O)(O)=O.CCOC(=O)C1=C[C@@H](OC(CC)CC)[C@H](NC(C)=O)[C@@H](N)C1

InChI

1S/C16H28N2O4.H3O4P/c1-5-12(6-2)22-14-9-11(16(20)21-7-3)8-13(17)15(14)18-10(4)19;1-5(2,3)4/h9,12-15H,5-8,17H2,1-4H3,(H,18,19);(H3,1,2,3,4)/t13-,14+,15+;/m0./s1

InChI key

PGZUMBJQJWIWGJ-ONAKXNSWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oseltamivir phosphate (impurity B free) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Henju Marjuki et al.
The Journal of infectious diseases, 210(3), 435-440 (2014-02-27)
Human infections caused by avian influenza A virus type subtype H7N9 have been associated with substantial morbidity and mortality. Emergence of virus variants carrying markers of decreased susceptibility to neuraminidase inhibitors was reported. Here we show that DAS181 (Fludase), an
Ding Yuan Oh et al.
The Journal of antimicrobial chemotherapy, 69(9), 2458-2469 (2014-05-21)
The emergence of the pandemic influenza A(H1N1)pdm09 virus in 2009 saw a significant increase in the therapeutic and prophylactic use of neuraminidase inhibitors (NAIs) to mitigate the impact of this highly transmissible virus. Prior to the pandemic, many countries stockpiled
Henju Marjuki et al.
Journal of virology, 89(10), 5419-5426 (2015-03-06)
Human infections by avian influenza A(H7N9) virus entail substantial morbidity and mortality. Treatment of infected patients with the neuraminidase (NA) inhibitor oseltamivir was associated with emergence of viruses carrying NA substitutions. In the NA inhibition (NI) assay, R292K conferred highly
Isabelle Marois et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(3), 973-987 (2014-11-22)
The clinical benefits of oseltamivir (Tamiflu) are well established, but the effects of antiviral treatment on the immune response are poorly understood. By use of flow cytometric analyses and the mouse model, we thoroughly investigated the impact of such a
Aihua Song et al.
Antiviral research, 111, 60-68 (2014-09-15)
Influenza virus is a global health concern due to its unpredictable pandemic potential. Frequent mutations of surface molecules, hemagglutinin (HA) and neuraminidase (NA), contribute to low efficacy of the annual flu vaccine and therapeutic resistance to standard antiviral agents. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service