Skip to Content
Merck
All Photos(1)

Key Documents

1.12537

Supelco

UV lamp 254 nm

suitable for thin layer chromatography (TLC)

Synonym(s):

254nm Ultraviolet Light

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
39101616
NACRES:
NB.21
Pricing and availability is not currently available.

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

no temp limit

General description

Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.
for thin-layer chromatography

Application



  • MassSpecPreppy-An end-to-end solution for automated protein concentration determination and flexible sample digestion for proteomics applications.: This research introduces an automated solution for protein concentration determination and sample digestion in proteomics, utilizing vials for precise and contamination-free sample handling (Reder et al., 2024).




Linkage

Replaces: 12537-1; 12537

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–6 of 6 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is this lamp suitable for a 8x8x0,1cm nylon membrane? It is needed for UV-crosslinking of RNA on the nylon membrane. Thank you

    1 answer
    1. The dimensions of this lamp are 16.1 x 2.4 x 5.5 cm, so the area of an 8 x 8 x 0.1 cm nylon membrane would not be fully covered for completely even irradiation. This lamp was developed specifically for use in visualizing thin-layer chromatography plates. It cannot be guaranteed to supply the power necessary for RNA crosslinking. Use of the product for this purpose is outside the original scope of design and falls under the discretion of the customer.

      Helpful?

  4. Buongiorno, volevo chiederle se il prodotto "1.12537" sia solo la lampada UV oppure tutto l'apparato con anche "il sistema elettrico" che permette di accendere e mettere in funzione la lampada. Mi potrebbe aiutare a capire? La ringrazio

    1 answer
    1. This is a whole electrical assembly with a UV lamp. It is mainly used for Thin Layer Chromatography analytical purposes.

      Helpful?

  5. IS THE BULB REPLACEABLE AVAILABLE ? if so, partnumber ?

    1 answer
    1. Unfortunately, a replacement bulb is not offered for this product. The UV lamp is guaranteed for six months from the date of delivery.

      Helpful?

  6. Is the bulb replaceable? Available where? What is its life expectancy?

    1 answer
    1. Unfortunately, a replacement bulb is not offered for this product. The UV lamp is guaranteed for six months from the date of delivery.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service