Skip to Content
Merck
All Photos(1)

Key Documents

Y0001237

Nimesulide for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Nimesulide, N-(4-Nitro-2-phenoxyphenyl)methanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N2O5S
CAS Number:
Molecular Weight:
308.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nimesulide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CS(NC1=C(OC2=CC=CC=C2)C=C([N+]([O-])=O)C=C1)(=O)=O

InChI

1S/C13H12N2O5S/c1-21(18,19)14-12-8-7-10(15(16)17)9-13(12)20-11-5-3-2-4-6-11/h2-9,14H,1H3

InChI key

HYWYRSMBCFDLJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nimesulide for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Highly selective cyclooxygenase-2 inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Suleyman et al.
Current medicinal chemistry, 15(3), 278-283 (2008-03-13)
In this review it is shown that nimesulide, a selective cyclooxigenase-2 (COX-2) inhibitor, is different from other selective COX-2 inhibitors and classical non-steroidal anti-inflammatory drugs (NSAIDs). The anti-inflammatory effect mechanism of nimesulide (inhibition of inflammatory mediators) is similar to other
Lorena Pochini et al.
Biochemical pharmacology, 89(3), 422-430 (2014-04-08)
The effect of pharmaceutical compounds on the rat kidney B0AT1 transporter in proteoliposomes has been screened. To this aim, inhibition of the transport activity by the different compounds was measured on Na(+)-[(3)H]glutamine co-transport in the presence of membrane potential positive
M Page et al.
Annales francaises d'anesthesie et de reanimation, 27(9), 742-746 (2008-09-02)
Nimesulide is a non-steroidal anti-inflammatory drug available in several European countries. A hepatic toxicity due to nimesulide has been reported but fatal cases remain rare. We report the case of a 49-year-old woman treated by nimesulide during three days, admitted
E C Huskisson
Clinical and experimental rheumatology, 19(1 Suppl 22), S21-S25 (2001-04-12)
World-wide experience with nimesulide confirms that it is an effective anti-inflammatory drug in the treatment of osteoarthritis. A review of several studies in this condition confirms that nimesulide is at least as efficacious as other commonly used compounds. The safety
K D Rainsford
Current medical research and opinion, 22(6), 1161-1170 (2006-07-19)
This paper summarises the outcome from a consensus meeting, held in Rome on 5 October 2005, that aimed to review the state of the art regarding the non-steroidal anti-inflammatory drug (NSAID) nimesulide, with reference to its chemical, pharmacokinetic, pharmacological and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service