Merck
Nimesulide, a balanced drug for the treatment of osteoarthritis.
Clinical and experimental rheumatology (2001-04-12)
E C Huskisson
ABSTRACT
World-wide experience with nimesulide confirms that it is an effective anti-inflammatory drug in the treatment of osteoarthritis. A review of several studies in this condition confirms that nimesulide is at least as efficacious as other commonly used compounds. The safety profile of nimesulide, compared to reference drugs such as naproxen, etodolac and diclofenac, demonstrates superior gastrointestinal tolerability. Nimesulide is therefore a good choice for the long-term treatment of OA.
Product Description
Supelco
Nimesulide, Pharmaceutical Secondary Standard; Certified Reference Material
Nimesulide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nimesulide
Nimesulide for peak identification, European Pharmacopoeia (EP) Reference Standard