Skip to Content
Merck
All Photos(1)

Documents

Y0001113

Triolein

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glyceryl trioleate, (9Z)9-Octadecenoic acid 1,2,3-propanetriyl ester, 1,2,3-Tri(cis-9-octadecenoyl)glycerol, Glycerol trioleate, Glycerol triolein, Oleic acid triglyceride, Oleic triglyceride, TG(18:1(9Z)/18:1(9Z)/18:1(9Z)), Triolein

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H33COOCH2)2CHOCOC17H33
CAS Number:
Molecular Weight:
885.43
Beilstein:
1718692
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triolein

manufacturer/tradename

EDQM

bp

235-240 °C/18 mmHg (lit.)

density

0.91 g/mL (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[H]C(COC(CCCCCCC/C=C\CCCCCCCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-

InChI key

PHYFQTYBJUILEZ-IUPFWZBJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triolein EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

626.0 °F - closed cup

Flash Point(C)

330.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Muth et al.
Colloids and surfaces. B, Biointerfaces, 149, 280-287 (2016-10-23)
Lipase-catalyzed hydrolysis of triglycerides yields glycerol and free fatty-acids, provided that the enzyme is non-regioselective. For an Sn-1,3 regioselective enzyme, such as lipase from Thermomyces lanuginosus, the final product is no longer glycerol but Sn-2 monoglyceride instead. However, surface active
Thalita Dallapícula Ramos et al.
Analytical and bioanalytical chemistry, 408(29), 8495-8503 (2016-10-14)
The versatile, easy, and rapid atmospheric monitor (VERAM), a passive sampler device widely used for air monitoring, was evaluated as passive sampler for the determination of 23 pesticides in water. Gas chromatography with mass spectrometry detection was employed for determination
Ronit Mesilati-Stahy et al.
PloS one, 13(12), e0209565-e0209565 (2019-01-01)
The effects of the macrostructure of long chain fatty acids on the lipid metabolism and biosynthesis of lipid droplets (LD) was studied in mammary epithelial cells (MEC). MEC were exposed to similar compositions and concentrations of fatty acids in the
Lili Xiong et al.
Cell transplantation, 28(6), 704-712 (2018-11-20)
Fat embolism syndrome (FES) is a common complication following long bone fracture; fat droplets are released into the blood circulation and form embolisms, mainly in lung and brain. However, the potential mechanisms involved remain to be clarified. In this study
Ouma Cissé et al.
European journal of nutrition, 58(6), 2411-2423 (2018-09-01)
Poor maternal nutrition sensitises to the development of metabolic diseases and obesity in adulthood over several generations. The prevalence increases when offspring is fed with a high-fat (HF) diet after weaning. This study aims to determine whether such metabolic profiles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service