Skip to Content
Merck
All Photos(5)

Key Documents

T5888

Sigma-Aldrich

Glyceryl tripalmitate

≥99%

Synonym(s):

TG(16:0/16:0/16:0), Glycerol tripalmitate, Tripalmitin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)14COOCH2]2CHOCO(CH2)14CH3
CAS Number:
Molecular Weight:
807.32
Beilstein:
1811188
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

semisynthetic

Assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3

InChI key

PVNIQBQSYATKKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glyceryl tripalmitate is a simple saturated glyceride. It is a solid present majorly in palm and coconut oil. It constitutes 1-11% of palm oil and has high melting temperature.

Application

Glyceryl tripalmitate has been used as a reference standard
  • for the generation of calibration curve generation for the quantification of lipids from Azospirillum brasilense
  • for the quantification of silk worm embryos and extraembryonic yolk lipids using thin layer chromatography
  • for the quantification of triacylglycerol in Neochloris minuta microalgal cells

Biochem/physiol Actions

Glyceryl tripalmitate is a carrier molecule for effective drug delivery and is used solid lipid nanoparticle formulations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changes in lipid droplet localization during embryogenesis of the silkworm, Bombyx mori
Yamahama Y, et al.
Zoological Science, 25(6), 580-587 (2008)
Isothermal crystallization of tripalmitin in sesame oil
Dibildox-Alvarado E and Toro-Vazquez JF
Journal of the American Oil Chemists' Society, 74(2), 69-76 (1997)
Tripalmitin nanoparticle formulations significantly enhance paclitaxel antitumor activity against breast and lung cancer cells in vitro
Leiva M, et al.
Scientific reports, 7(1), 13506-13506 (2017)
Starch, Lipid, and Protein Accumulation in Nutrient-Stressed Microalgal Cells Studied Using Fourier Transform Infrared Microscopy
Hastings G, et al.
Frontiers in Science, Technology, Engineering and Mathematics, 2(2), 1-1 (2018)
Remington: The Science and Practice of Pharmacy, 1 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service