Skip to Content
Merck
All Photos(1)

Key Documents

Y0000538

Dexamethasone isonicotinate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dexamethasone 21-isonicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32FNO6
CAS Number:
Molecular Weight:
497.56
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dexamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

F[C@@]21[C@H]([C@H]4[C@@]([C@@]([C@@H](C4)C)(O)C(=O)COC(=O)c5ccncc5)(C[C@@H]2O)C)CCC3=CC(=O)C=C[C@@]31C

InChI

1S/C28H32FNO6/c1-16-12-21-20-5-4-18-13-19(31)6-9-25(18,2)27(20,29)22(32)14-26(21,3)28(16,35)23(33)15-36-24(34)17-7-10-30-11-8-17/h6-11,13,16,20-22,32,35H,4-5,12,14-15H2,1-3H3/t16-,20+,21+,22+,25+,26+,27+,28+/m1/s1

InChI key

BQTXJHAJMDGOFI-NJLPOHDGSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone isonicotinate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A C Moreira et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 24(7), 339-341 (1992-07-01)
A previous study in an untreated girl with 17 alpha-hydroxylase deficiency syndrome (17-OHDS) indicated that in the absence of cortisol production, ACTH and corticosterone (B) may interact to modulate the brain-pituitary-adrenocortical system. The ACTH-B interplay was further examined in this
N E Robinson et al.
Equine veterinary journal, 34(1), 17-22 (2002-01-31)
This study used a cross-over design to compare the efficacy of 3 corticosteroids for the relief of airway obstruction and inflammation in 9 heaves-affected horses. The severity of airway obstruction and inflammation was quantified by measurement of lung function and
Jorge Elizondo-Rodriguez et al.
Foot & ankle international, 34(1), 8-14 (2013-02-07)
The objective of this study was to compare intramuscularly applied botulinum toxin A (BTX-A) in the gastroc-soleus complex with intralesional steroids for the treatment of plantar fasciitis. The patients were randomly divided into 2 groups according to the treatment received.
M Kusenda et al.
Journal of veterinary internal medicine, 27(1), 200-206 (2012-12-01)
Dexamethasone frequently is used for treatment of ketosis in dairy cows, but its effects are not fully understood. Dexamethasone treatment affects whole body insulin sensitivity. Twelve German Holstein cows, 2-4 weeks postpartum, 5 days after omentopexy to correct left abomasal
[Notes on the disaster in Bhopal].
W F Diller
Deutsche medizinische Wochenschrift (1946), 110(45), 1749-1751 (1985-11-08)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service