Skip to Content
Merck
All Photos(1)

Key Documents

Y0000235

Almagate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

almagate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Al](O)(O)O.[Mg+2].[Mg+2].[Mg+2].[O-]C(=O)[O-].[O-H].[O-H].[O-H].[O-H].O.O

InChI

1S/CH2O3.Al.3Mg.9H2O/c2-1(3)4;;;;;;;;;;;;;/h(H2,2,3,4);;;;;9*1H2/q;+3;3*+2;;;;;;;;;/p-9

InChI key

MTEOMEWVDVPTNN-UHFFFAOYSA-E

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Almagate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Sárszegi et al.
Orvosi hetilap, 141(35), 1915-1917 (2000-10-06)
After administration of a single dose of an aluminium containing antacid (Tisacid) to eight non dialysis patients in various stages of chronic renal insufficiency, the kinetics of serum aluminium absorption was studied. Serum aluminium peaked at two points of time
S Massarrat et al.
Gut, 29(3), 291-297 (1988-03-01)
In order to identify the risk factors affecting the healing of duodenal ulcer, a clinical trial with effective dose of antacid was carried out in 53 patients. Duration of ulcer history, number of relapses, duration of the last and present
T Escalera Temprado et al.
Anales de medicina interna (Madrid, Spain : 1984), 21(2), 81-83 (2004-02-21)
Nowadays, the human immunodeficiency virus infection (HIV) is a chronic disease. In the frequent clinical situations with fever, lymph nodes and loss weight it is necessary to determine their etiology, for establishing a specific treatment. Gastrointestinal opportunistic infections or gastric
L Grande Posa et al.
Revista espanola de enfermedades digestivas : organo oficial de la Sociedad Espanola de Patologia Digestiva, 83(4), 229-234 (1993-04-01)
The effect of postprandial administration of a new formulation of almagate (1.5 g, po) or an association of alginate + antacid (2.4 g, po) on gastric alkalinization and gastroesophageal reflux were assessed in a randomized, single-blind, cross-over study in 21
[The prescription of almagate as an indicator of quality in medical performance].
A Jimeno et al.
Atencion primaria, 9(6), 339-340 (1992-04-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service