Skip to Content
Merck
All Photos(3)

Key Documents

C1629

Sigma-Aldrich

Choline bitartrate

Synonym(s):

2-(Hydroxyethyl)trimethylammonium bitartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO7
CAS Number:
Molecular Weight:
253.25
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

SMILES string

[O-]C([C@H](O)[C@@H](O)C(O)=O)=O.C[N+](C)(C)CCO

InChI

1S/C5H14NO.C4H6O6/c1-6(2,3)4-5-7;5-1(3(7)8)2(6)4(9)10/h7H,4-5H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/q+1;/p-1/t;1-,2-/m.1/s1

InChI key

QWJSAWXRUVVRLH-LREBCSMRSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Choline bitartrate has been used as one of the components in experimental diets to study the effects on reproductive processes and on bone genes during gestation and lactation.

Biochem/physiol Actions

Acyl group acceptor
Choline bitartrate is known to serve as a dietary supplement. Choline acts as a precursor for acetylcholine synthesis.

Substrates

Choline acetyltransferase substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extra virgin olive oil in maternal diet increases osteogenic genes expression, but high amounts have deleterious effects on bones in mice offspring at adolescence
Mousavi SN, et al.
Iranian Journal of Basic Medical Sciences, 19(12), 1299-1299 (2016)
Siamak Sheikhi et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 84(2), 109-121 (2024-02-05)
The mother's thyroid hormone status during gestation and the first few months after delivery can play a crucial role in maturation during the brain development of the child. Transient abnormalities in thyroid function at birth indicate developmental and cognitive disorders
Effects of Diets Enriched in Omega-9 or Omega-6 Fatty Acids on Reproductive Process
Mousavi SN, et al.
Journal of family & reproductive health, 10(2), 85-85 (2016)
No acute effects of choline bitartrate food supplements on memory in healthy, young, human adults
Lippelt DP, et al.
PLoS ONE, 11(6), e0157714-e0157714 (2016)
Margaret A Mouat et al.
Cells, 11(11) (2022-06-11)
Understanding the neurogenic causes of obesity may reveal novel drug targets to counter the obesity crisis and associated sequelae. Here, we investigate whether the deletion of GPR37L1, an astrocyte-specific orphan G protein-coupled receptor, affects whole-body energy homeostasis in mice. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service