Skip to Content
Merck
All Photos(1)

Key Documents

PHR1364

Supelco

Allantoin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Allantoin, 5-Ureidohydantoin, Glyoxylic(acid) diureide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3
CAS Number:
Molecular Weight:
158.12
Beilstein:
102364
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. A0349000
traceable to USP 1012939

API family

allantoin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

230 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NC(=O)NC1NC(=O)NC1=O

InChI

1S/C4H6N4O3/c5-3(10)6-1-2(9)8-4(11)7-1/h1H,(H3,5,6,10)(H2,7,8,9,11)

InChI key

POJWUDADGALRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allantoin, the final catabolic product of purines in mammals,[1] is a highly polar and amphoteric substance.[2] Allantoin is mainly used for skin protection as the drug promotes cell proliferation and wound healing.[3]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Allantoin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Purine metabolite via the uric acid pathway. Uric acid also reacts with free radicals to produce allantoin, thus allantoin may be a useful biomarker for oxidative stress.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1468 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quantification of allantoin in various Zea mays L. hybrids by RP-HPLC with UV detection
Maksimovic K, et al.
Pharmazie, 59(7), 524-527 (2004)
Simultaneous determination of allantoin and glycolic acid in snail mucus and cosmetic creams with high performance liquid chromatography and ultraviolet detection
El Mubarak MAS, et al.
Journal of Chromatography A, 1322, 49-53 (2013)
Simultaneous quantification of allantoin and steroidal saponins in yam (Dioscorea spp.) powders
Maksimovic K, et al.
Journal of Applied Research on Medicinal and Aromatic Plants, 13(7), 100200-100200 (2019)
[Allantoin].
T Ogihara et al.
Nihon rinsho. Japanese journal of clinical medicine, 57 Suppl, 769-771 (2001-02-07)
Allantoin degradation by Saccharomyces cerevisiae--a model system for gene regulation and metabolic integration.
T G Cooper
Advances in enzymology and related areas of molecular biology, 56, 91-139 (1984-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service