Skip to Content
Merck
All Photos(2)

Documents

B6906

Sigma-Aldrich

Aloin

from Aloe barbadensis Miller leaves, ≥97%

Synonym(s):

1,8-Dihydroxy-10-(β-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenone, 10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone, Aloin A, Barbaloin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O9
CAS Number:
Molecular Weight:
418.39
Beilstein:
6077558
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

Aloe barbadensis Miller leaves

Assay

≥97%

form

powder

solubility

methanol: 100 mg/mL, clear to very slightly hazy, light yellow to yellow

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@H]2c3cccc(O)c3C(=O)c4c(O)cc(CO)cc24

InChI

1S/C21H22O9/c22-6-8-4-10-14(21-20(29)19(28)17(26)13(7-23)30-21)9-2-1-3-11(24)15(9)18(27)16(10)12(25)5-8/h1-5,13-14,17,19-26,28-29H,6-7H2/t13-,14+,17-,19+,20-,21+/m1/s1

InChI key

AFHJQYHRLPMKHU-OSYMLPPYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aloin, a natural mixed diastereomer (A/B) anthracycline from aloe plant, may be studied as a chemopreventive factor and antineoplastic agent. Aloin inhibits angiogenesis, enhances melanogenesis and regulates transglutaminase activity. Aloin may be used to assess its activity as a selective phosphodiesterase inhibitor. Aloin A is used as a stabilizer in the preparation of gold and silver nanoparticles.

Biochem/physiol Actions

Purgative activity in rats requires activation by Eubacterium sp. strain BAR or similar intestinal anaerobe, presumably by conversion to aloe-emodin anthrone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeljka Krpetić et al.
Langmuir : the ACS journal of surfaces and colloids, 25(13), 7217-7221 (2009-06-10)
A novel use of two components of Cape aloe, aloin A and aloesin, acting as stabilizers in the preparation of gold and silver nanoparticles, is reported. Stable water-soluble particles of different size and shape are prepared by varying the reaction
Aloin enhances cisplatin antineoplastic activity in B16-F10 melanoma cells by transglutaminase-induced differentiation.
Tabolacci C, Rossi S, Lentini A, et al.
Amino Acids, doi: 10-doi: 10 (2011)
Angelo Gismondi et al.
Amino acids, 38(1), 257-262 (2009-02-13)
The aim of this study was to collect evidences on the role of transglutaminase (TG, E.C.2.3.2.13) in the antineoplastic properties exerted by nimesulide (NMS), a non-steroidal anti-inflammatory drug, on murine B16-F10 melanoma cells. Treatment of melanoma cells with nimesulide produces
Mahmoud A Elsohly et al.
Journal of AOAC International, 90(1), 28-42 (2007-03-22)
Methods using liquid chromatography/mass spectrometry (LC/MS) and LC with diode array detection (DAD) in the UV range (LC/UV) were developed for the determination of low levels of the anthraquinones aloe-emodin and aloin-A (barbaloin) in aloe-based products. The methods were used
Claudio Tabolacci et al.
Life sciences, 87(9-10), 316-324 (2010-07-14)
Aloe-emodin (AE), a natural hydroxyanthraquinone compound, has been reported as a potential anticancer agent. We studied the antineoplastic properties of AE on highly metastatic B16-F10 melanoma murine cells. Cell proliferation was assessed by cell counting and viability was investigated using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service