Skip to Content
Merck
All Photos(1)

Key Documents

PHR1282

Supelco

Azathioprine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

6-[(1-Methyl-4-nitro-1H imidazol-5yl) sulfanyl]-7H-purine, 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N7O2S
CAS Number:
Molecular Weight:
277.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 26
traceable to Ph. Eur. A1500000
traceable to USP 1046001

API family

azathioprine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cn1cnc(c1Sc2ncnc3[nH]cnc23)[N+]([O-])=O

InChI

1S/C9H7N7O2S/c1-15-4-14-7(16(17)18)9(15)19-8-5-6(11-2-10-5)12-3-13-8/h2-4H,1H3,(H,10,11,12,13)

InChI key

LMEKQMALGUDUQG-UHFFFAOYSA-N

Gene Information

human ... PPAT(5471)

Looking for similar products? Visit Product Comparison Guide

General description

Azathioprine is a potentially useful antileukaemic drug, with immunosuppressive action, which is administered orally or by I.V route. It is widely used to treat a variety of auto-immune disorders and also has a marked effect on T-lymphocytes and suppresses cell mediated immunity.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Azathioprine may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using 1H NMR spectroscopy and spectrophotometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0017 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quantitative determination of azathioprine in tablets by 1H NMR spectroscopy
Goger GN, et al.
Journal of Pharmaceutical and Biomedical Analysis, 21(3), 685-690 (2000)
Spectrophotometric determination of azathioprine in bulk and pharmaceutical dosage forms
Bhaskar M, et al.
International Journal of ChemTech Research, 2(1), 376-378 (2010)
Systemic therapies for severe atopic dermatitis in children and adults.
Carsten Flohr et al.
The Journal of allergy and clinical immunology, 132(3), 774-774 (2013-09-03)
Loïc Guillevin et al.
The New England journal of medicine, 371(19), 1771-1780 (2014-11-06)
The combination of cyclophosphamide and glucocorticoids leads to remission in most patients with antineutrophil cytoplasm antibody (ANCA)-associated vasculitides. However, even when patients receive maintenance treatment with azathioprine or methotrexate, the relapse rate remains high. Rituximab may help to maintain remission.
Björn Pasternak et al.
American journal of epidemiology, 177(11), 1296-1305 (2013-03-22)
Increased risks of lymphoma and skin cancer associated with thiopurine use among patients with inflammatory bowel disease have been shown, but data on the overall cancer risk are limited. We conducted a historical cohort study of 45,986 patients with inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service