Skip to Content
Merck
All Photos(1)

Key Documents

P9422

Supelco

Pramoxine hydrochloride

analytical standard, ≥98%

Synonym(s):

4-[3-(4-Butoxyphenoxy)propyl]morpholine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO3 · HCl
CAS Number:
Molecular Weight:
329.86
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CCCCOc1ccc(OCCCN2CCOCC2)cc1

InChI

1S/C17H27NO3.ClH/c1-2-3-12-20-16-5-7-17(8-6-16)21-13-4-9-18-10-14-19-15-11-18;/h5-8H,2-4,9-15H2,1H3;1H

InChI key

SYCBXBCPLUFJID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pramoxine hydrochloride is a surface and local anesthetic agent , which provides relief from itching and pain for a short period of time.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Topical anesthetic for skin or mucous membranes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzana Ljubojević et al.
Acta dermatovenerologica Croatica : ADC, 16(2), 96-100 (2008-06-11)
Perioral dermatitis is an inflammatory facial skin disorder that predominantly affects women. It is rarely diagnosed in children. The etiology of perioral dermatitis is unknown; however, uncritical use of topical corticosteroids often precedes skin lesions. There is a written diagnostic
Trudye A Young et al.
The Journal of dermatological treatment, 20(2), 76-81 (2008-09-30)
The objective of this study was to evaluate the efficacy of a commercially available anti-itch lotion containing 1% pramoxine hydrochloride versus control lotion in the treatment of uremic pruritus in adult hemodialysis patients. This was a randomized, double-blind, controlled comparative
OTC product: Monistat Soothing Care for vaginial discomfort.
Seena L Zierler-Brown
Journal of the American Pharmacists Association : JAPhA, 45(6), 768-768 (2005-12-31)
The safety of pramoxine hydrochloride when used as a topical (surface) anesthetic.
A A Fisher
Cutis, 62(3), 122-123 (1998-10-14)
Contact dermatitis from prilocaine with cross-sensitivity to pramocaine and bupivacaine.
Fernando García et al.
Contact dermatitis, 56(2), 120-121 (2007-01-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service