NIST1963A
Nominal 100 nm diameter polystyrene spheres
NIST® SRM® 1963a
Synonym(s):
Polystyrene spheres
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
Recommended Products
form
suspension
packaging
pkg of 5 mL
manufacturer/tradename
NIST®
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service