Skip to Content
Merck
All Photos(2)

Key Documents

NIST1898

Titanium dioxide

NIST® SRM® 1898, nanomaterial

Synonym(s):

Titanium(IV) oxide, Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 15 g

manufacturer/tradename

NIST®

mp

>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 1898 contains an amber glass bottle with approximately 15 g of anatase and rutile mixed phasenanocrystalline titanium dioxide (TiO2) in a dry agglomerated powder form.

SRM 1898_cert

SRM 1898_SDS

Features and Benefits

  • The certified values for multi-point (MP) and single-point(SP) data analysis using the linear form of the single parameter BET equationare presented and evaluated for the Brunauer-Emmet-Teller (BET) specificsurface area, determined through nitrogen gas sorption at liquid nitrogentemperature (77.3 K) using the discontinuous manometric measurement method, inaccordance with ISO 9277:2010.
  • Reference value is provided for nicotine adeninedinucleotide hydrate equivalent to photocatalytic activity.
  • Elemental composition, purity,crystalline-phase content, and crystallite size information values are provided for both Anatase and Rutile.
  • Expiration detailsand instructions for storage, handling, use, and maintenance are provided on the NIST certificate.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is this a mix of anatase and rutile ?

    1 answer
    1. This product is supplied by NIST. Please see pages 5 and 6 of the certificate/report which explains the mix of anatase and rutile:
      https://tsapps.nist.gov/srmext/certificates/1898.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service