Skip to Content
Merck
All Photos(1)

Documents

M5651

Supelco

Mepenzolate bromide

analytical standard

Synonym(s):

3-Hydroxy-1,1-dimethylpiperidinium bromide benzilate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H26NO3Br
CAS Number:
Molecular Weight:
420.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Br.C[N]1(C)CCCC(C1)OC(=O)C(O)(c2ccccc2)c3ccccc3

InChI

1S/C21H26NO3.BrH/c1-22(2)15-9-14-19(16-22)25-20(23)21(24,17-10-5-3-6-11-17)18-12-7-4-8-13-18;/h3-8,10-13,19,24H,9,14-16H2,1-2H3;1H

InChI key

FOPUTUUUOOOXQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R F Ochillo et al.
Research communications in chemical pathology and pharmacology, 36(3), 503-506 (1982-06-01)
The mydriatic effects of mepenzolate, a quarternary antimuscarinic agent, has been investigated and compared with those of atropine by pupillographic analysis in rabbits. The time-response curve for the onset of action for the agents (half-time, ca.5 min) was the same
G R Murray et al.
Journal of chromatography, 308, 143-151 (1984-06-08)
A new sensitive and selective capillary column gas chromatographic method for the anti-cholinergic agent glycopyrronium bromide in human plasma is described. The procedure involves preliminary ion-pair extraction of the drug into dichloromethane, followed by concentration and analysis of the ion-pair
M Cohen-Armon et al.
Journal of cardiovascular pharmacology, 6(6), 1148-1155 (1984-11-01)
The possible interaction between the muscarinic receptor and the antiarrythmic drug amiodarone was studied physiologically in the guinea pig ileum, as well as by competition binding experiments in rat brain and cardiac tissues, using the highly specific tritiated muscarinic antagonist
C S Tsai et al.
Journal of ethnopharmacology, 45(3), 215-222 (1995-03-01)
In our attempt to isolate the pharmacologically active ingredients in the aqueous extracts of Mareya micrantha, we have selected the contractions of the longitudinal muscle of the isolated guinea-pig ileum preparation as a pharmacological marker to monitor retention of pharmacological
C S Tsai et al.
Research communications in chemical pathology and pharmacology, 55(2), 275-278 (1987-02-01)
The responsiveness of the cholinergic receptor (ChR) of the longitudinal muscle of guinea-pig isolated ileum was characterized by determining the ED50 of acetylcholine (ACh) and pA2 of mepenzolate (MPZ) in tissues from animals of different ages. The ages of guinea

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service