Skip to Content
Merck
All Photos(2)

Documents

L2378

Sigma-Aldrich

γ-Linolenic acid

≥99%, liquid

Synonym(s):

gamma-Linolenic acid, cis,cis,cis-6,9,12-Octadecatrienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H30O2
CAS Number:
Molecular Weight:
278.43
Beilstein:
1712253
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant oil (borage)

Assay

≥99%

form

liquid

functional group

carboxylic acid

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O

InChI

1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10,12-13H,2-5,8,11,14-17H2,1H3,(H,19,20)/b7-6-,10-9-,13-12-

InChI key

VZCCETWTMQHEPK-QNEBEIHSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

γ-linolenic acid has been used as an analytical standard in gas chromatography. It may be used in nutritional studies regarding weight regain and as a possible tumor suppression agent. γ-linolenic acid is used in studies on the mechanisms and prevention of oxidation/peroxidation of unsaturated fatty acids.

Biochem/physiol Actions

γ-Linolenic acid , a polyunsaturated fatty acid, exerts anti-inflammatory effect by decreasing both interleukin (IL) 6 and IL-8 production in the human enterocyte-like cell line Caco-2 and humandendritic cells in vitro .
Gamma-linolenate (C18:6,9,12) differs from α-linolenate (C18:9,12,15) in the positions of the double bonds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julianne Lindemann et al.
Current medical research and opinion, 25(12), 2865-2875 (2009-10-09)
Asthma can have a negative impact on quality of life although this is not well correlated with objective evaluations of pulmonary function. A medical food, EFF1009, containing the fatty acids gamma-linolenic acid (GLA) and eicosapentaenoic acid (EPA) decreases leukotriene B(4)
J Hindo et al.
Radiation research, 153(2), 201-207 (2000-01-12)
Peroxidation of unconjugated polyunsaturated fatty acids such as linolenic acid proceeds through a free radical chain mechanism and is accompanied by the formation of conjugated dienes such as hydroperoxides. In an investigation of radiation-induced oxidation of aqueous linolenate, we have
P Kankaanpää et al.
Allergy, 56(7), 633-638 (2001-06-26)
The increased consumption of n-6 polyunsaturated fatty acids (PUFA) has been shown to coincide with the increased prevalence of atopic diseases. We aimed to investigate whether maternal diet and atopic status influence the PUFA composition of breast milk and the
R J Levy et al.
Clinical pharmacology and therapeutics, 25(5 Pt 1), 562-570 (1979-05-01)
The urinary excretion of gamma-carboxyglutamic acid (Gla), the amino acid involved in the vitamin K-dependent calcium binding of prothrombin and clotting factors VII, IX, and X, was studied in warfarin-anticoagulated patients. An isotope dilution procedure was developed for the measurement
Nutritional and medical importance of gamma-linolenic acid.
D F Horrobin
Progress in lipid research, 31(2), 163-194 (1992-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service